EMAIL THIS PAGE TO A FRIEND
Journal of cranio-maxillo-facial surgery : official publication of the European Association for Cranio-Maxillo-Facial Surgery
Influence of epidermal growth factor receptor expression on the cetuximab and panitumumab response rates of head and neck carcinoma cells.
PMID 24780353
Abstract
To examine the impact of epidermal growth factor receptor (EGFR) expression level on the efficacy of monoclonal antibodies against the EGFR. In four human head and neck carcinoma cell lines, epidermal growth factor expression was knocked down by lentiviral RNA interference. Next, the efficacies of cetuximab and panitumumab at concentrations of 4, 40, and 400 μg/ml were measured by real time cell analysis for a 48-h duration. Finally, the different response rates to the drugs were statistically analyzed. The lentiviral EGFR knockdown efficiency ranged from 18 to 54 % across all of the cell lines. All original cell lines exhibited rather poor or inverse responses with regard to EGFR-AB treatment. In contrast, inhibiting EGFR expression in the same cell lines yielded statistically significant better responses to cetuximab or panitumumab treatment. The cell lines used in this study responded poorly to cetuximab and panitumumab. Better anti-EGFR treatment efficacy was related to lower EGFR expression in head and neck cancer cell lines. These findings might influence the selection of patients to receive cetuximab and panitumumab treatment for head and neck cancer.