EMAIL THIS PAGE TO A FRIEND
Journal of molecular endocrinology
The INSL3 gene is a direct target for the orphan nuclear receptor, COUP-TFII, in Leydig cells.
PMID 24780841
Abstract
Insulin-like 3 (INSL3), a hormone produced by Leydig cells, regulates testicular descent during foetal life and bone metabolism in adults. Despite its importance, little is known about the molecular mechanisms controlling INSL3 expression. Reduced Insl3 mRNA levels were reported in the testis of mice deficient for chicken ovalbumin upstream promoter-transcription factor II (COUP-TFII), an orphan nuclear receptor known to play critical roles in cell differentiation and lineage determination in several tissues. Although COUP-TFII-deficient mice had Leydig cell dysfunction and impaired fertility, it remained unknown whether Insl3 expression was directly regulated by COUP-TFII. In this study, we observed a significant decrease in Insl3 mRNA levels in MA-10 Leydig cells depleted of COUP-TFII. Furthermore, a -1087 bp mouse Insl3 promoter was activated fourfold by COUP-TFII in MA-10 Leydig cells. Using 5' progressive deletions, the COUP-TFII-responsive element was located between -186 and -79 bp, a region containing previously uncharacterised direct repeat 0-like (DR0-like) and DR3 elements. The recruitment and direct binding of COUP-TFII to the DR0-like element were confirmed by chromatin immunoprecipitation and DNA precipitation assay respectively. Mutation of the DR0-like element, which prevented COUP-TFII binding, significantly decreased COUP-TFII-mediated activation of the -1087 bp Insl3 reporter in CV-1 fibroblast cells but not in MA-10 Leydig cells. Finally, we found that COUP-TFII cooperates with the nuclear receptor steroidogenic factor 1 (SF1) to further enhance Insl3 promoter activity. Our results identify Insl3 as a target for COUP-TFII in Leydig cells and revealed that COUP-TFII acts through protein-protein interactions with other DNA-bound transcription factors, including SF1, to activate Insl3 transcription in these cells.