EMAIL THIS PAGE TO A FRIEND
Annals of hematology
Correlation of CD11b and CD56 expression in adult acute myeloid leukemia with cytogenetic risk groups and prognosis.
PMID 24782118
Abstract
Among other phenotypic markers, CD11b expression has been considered as an unfavorable prognostic factor, both in terms of overall survival (OS), disease-free survival (DFS), and attainment and duration of complete remissions (CRs) in adult patients with acute myeloid leukemia (AML). Recently, some groups have restricted its prognostic impact to poor prognostic karyotypic risk groups. The aim of this study was to retrospectively analyze the prevalence of CD11b and of CD56 expression in blast cells of 158 AML patients [excluding those with t(15;17)] stratified according to their cytogenetic risk and to correlate these phenotypic characteristics with OS, DFS, and CR. CD11b was more frequently expressed in intermediate and unfavorable cytogenetic prognostic groups (38.9 and 35.5xa0%, respectively) than in the favorable group (9.5xa0%). No differences were observed in CD56 expression according to the cytogenetic risk groups. When OS, DFS, and CR were analyzed according to these two markers, no statistical differences were recorded in any cytogenetic risk group. In conclusion, although CD11b was more frequently expressed in blast cells of patients with intermediate and unfavorable cytogenetic risk groups, this feature did not translate into different clinical outcome. Similarly, CD56 positivity did not have any influence on the prognosis of these patients.