EMAIL THIS PAGE TO A FRIEND
Journal of feline medicine and surgery
Feline double pigtail ureteric stents for management of ureteric obstruction: short- and long-term follow-up of 26 cats.
PMID 24782460
Abstract
The objective of this study was to determine the outcome of cats with ureteric obstruction managed with double pigtail ureteric stents and to document the incidence of lower urinary tract signs at long-term follow-up. Data were obtained retrospectively from the medical records (2009-2012) of 26 cats that underwent ureteric stent placement. Owners were contacted for follow-up, and a quality of life questionnaire completed. Survival to discharge after stent placement was 85% (22/26). Prevalence of postoperative uroabdomen necessitating further surgery was 15% (4/26). Stents were replaced 4-28 months after the initial surgery in four cats because of migration, fracture, encrustation causing luminal obstruction or sterile cystitis, respectively. Nine cats were alive at follow-up, which was 3-28 months after the original surgery. Nine cats had azotaemic chronic kidney disease and nine had signs related to sterile cystitis; three of these cats were euthanased as a result of the severity of the signs. Preoperative serum creatinine of the survivors (9.4 mg/dl, n = 9) was not significantly different from that of the non-survivors (6.5 mg/dl, n = 13; P = 0.295). Quality of life was assigned a mean score of 8/10. Median survival of cats following discharge was 419 days (range 44-994 days). Signs consistent with sterile cystitis affected 35% of cats. It was concluded that ureteric stent placement in cats was associated with a 15% mortality rate before hospital discharge. Long-term management of ureteric stents is associated with a high rate of lower urinary tract signs.