EMAIL THIS PAGE TO A FRIEND
Drug development and industrial pharmacy
Deoxycholic acid-grafted PEGylated chitosan micelles for the delivery of mitomycin C.
PMID 24785368
Abstract
Mitomycin C (MTC) was incorporated to a micelle system preparing from a polymer named deoxycholic acid chitosan-grafted poly(ethylene glycol) methyl ether (mPEG-CS-DA). mPEG-CS-DA was synthesized and characterized by (1)H nuclear magnetic resonance ((1)H-NMR) and Fourier transform infrared spectroscopy. mPEG-CS-DA formed a core-shell micellar structure with a critical micelle concentration of 6.57 µg/mL. The mPEG-CS-DA micelles were spherical with a hydrodynamic diameter of about 231 nm. After poly(ethylene glycol)ylation of deoxycholic acid chitosan (CS-DA), the encapsulation efficiency and drug loading efficiency increased from 50.62% to 56.42% and from 20.51% to 24.13%, respectively. The mPEG-CS-DA micelles possessed a higher drug release rate than the CS-DA micelles. For pharmacokinetics, the area under the curve (AUC) of the mPEG-CS-DA micelles was 1.5 times higher than that of MTC injection, and these micelles can enhance the bioavailability of MTC. mPEG-CS-DA micelles reduced the distribution of MTC in almost all normal tissues and had the potential to improve the kidney toxicity caused by MTC injection.