EMAIL THIS PAGE TO A FRIEND
Protein engineering, design & selection : PEDS
Selection of high-affinity Centyrin FN3 domains from a simple library diversified at a combination of strand and loop positions.
PMID 24786107
Abstract
Alternative scaffold molecules represent a class of proteins important to the study of protein design and mechanisms of protein-protein interactions, as well as for the development of therapeutic proteins. Here, we describe the generation of a library built upon the framework of a consensus FN3 domain sequence resulting in binding proteins we call Centyrins. This new library employs diversified positions within the C-strand, CD-loop, F-strand and FG-loop of the FN3 domain. CIS display was used to select high-affinity Centyrin variants against three targets; c-MET, murine IL-17A and rat TNFα and scanning mutagenesis studies were used to define the positions of the library most important for target binding. Contributions from both the strand and loop positions were noted, although the pattern was different for each molecule. In addition, an affinity maturation schemexa0is described that resulted in a significant improvement in the affinity of one selected Centyrin variant. Together, this work provides important data contributing to our understanding of potential FN3 binding interfaces and a new tool for generating high-affinity scaffold molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
AV40493
Anti-RNMT antibody produced in rabbit, affinity isolated antibody
SAB2102036
Anti-RNMT antibody produced in rabbit, affinity isolated antibody
HPA039409
Anti-RNMT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
H9661 Hepatocyte Growth Factor human, HGF, recombinant, expressed in NSO cells, suitable for cell culture
H5791 Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
SRP6014 HGF human, recombinant, expressed in CHO cells, ≥97% (SDS-PAGE), ≥97% (HPLC)
SRP6166
HGF human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
RAB0116
Human GRO-α / CXCL1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0037
Human Procalcitonin ELISA Kit
WH0008731M1
Monoclonal Anti-RNMT antibody produced in mouse, clone 3H3-1D12, purified immunoglobulin, buffered aqueous solution
RAB0117
Mouse KC / CXCL1 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0118
Rat CINC-1 ELISA Kit, for serum, plasma and cell culture supernatant