EMAIL THIS PAGE TO A FRIEND
Natural product reports
Rocaglamide, silvestrol and structurally related bioactive compounds from Aglaia species.
PMID 24788392
Abstract
Covering: 2006 to 2013. Investigations on the chemistry and biology of rocaglamide, silvestrol and structurally related bioactive compounds from Aglaia species during the period 2006-2013 are reviewed. Included are new phytochemical studies of naturally occurring rocaglamide derivatives, an update on synthetic methods for cyclopenta[b]benzofurans, and a description of the recent biological evaluation and mechanism-of-action studies on compounds of this type.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0656 Rocaglamide, ≥96% (HPLC)
C29H31NO7