EMAIL THIS PAGE TO A FRIEND
Pharmaceutical research
Analgesic efficacy and safety of DALDA peptide analog delivery to the brain using oil-in-water nanoemulsion formulation.
PMID 24792826
Abstract
The main objective of this study was to develop and evaluate therapeutic efficacy and safety following systemic delivery of a peptide analgesic into the CNS using an oil-in-water nanoemulsion system. We have formulated a safe and effective, omega-3 rich polyunsaturated fatty acid containing oil-in-water nanoemulsion formulation, for encapsulating and delivering chemically-modified DALDA, a potent mu-opioid peptide analogue, to the CNS. One of the challenges with CNS delivery is the lack of a non-invasive bioanalytical technique to confirm CNS uptake and therapeutic efficacy. Using blood oxygen-level dependent (BOLD) functional magenetic resonance imaging (fMRI), we provide quantitative evidence of nanoemulsion-based delivery and analgesic activity of DALDA analogue in capsaicin-induced awake rat model of pain. Nanoemulsion formulation effectively encapsulated the modified analgesic peptide and demonstrated efficacy in the capsaicin- pain induced functional magnetic resonance imaging model in rodents. Preliminary safety evaluations show that the nanoemulsion system was well tolerated and did not cause any acute negative effects. Overall, these results show tremendous opportunity for the development of modified peptide analgesic-encapsulated nanoemulsion formulations for CNS delivery and therapeutic efficacy.