EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Simultaneous determination of two iridoid glycosides, two anthraquinones and four flavonoid glycosides of Zhi-Zi-Da-Huang decoction in rat plasma by UFLC-MS/MS: application to a comparative pharmacokinetic study in normal and cholestatic liver injury rats.
PMID 24793084
Abstract
A selective, sensitive and reliable ultra fast liquid chromatography-tandem mass spectrometry (UFLC-MS/MS) method has been developed for the simultaneous determination of two iridoid glycosides (geniposide and genipin gentiobioside), two anthraquinones (rhein and emodin) and four flavonoid glycosides (isonaringin, naringin, hesperidin and neohesperidin), the major active ingredients of Zhi-Zi-Da-Huang decoction (ZZDHD), in rat plasma using paeoniflorin as internal standard (IS). After liquid-liquid extraction with ethyl acetate-isopropanol (1:1, v/v), separation was achieved on a Shim-pack XR-ODS C18 column (75 mm×3.0 mm, 2.2 μm) using gradient elution with a mobile phase consisting of water (containing 0.1% formic acid) and acetonitrile at a flow rate of 0.4 mL/min. Detection was performed on 4000 QTRAP mass spectrometry equipped with turbo ion spray source in the negative ionization and multiple reaction monitoring (MRM) mode. The intra- and inter-day precisions (as relative standard deviation) were less than 11.4%, and accuracy (as relative error) was within ± 10.0%. The lower limits of quantification (LLOQ) were 4.0, 0.5, 2.0, 0.1, 1.0, 2.0, 1.0, 2.0 ng/mL for geniposide, genipin gentiobioside, rhein, emodin, isonaringin, naringin, hesperidin and neohesperidin, respectively. The extraction recoveries of the analytes and IS from rat plasma were all more than 86.0%. The method was fully validated and applied to compare the pharmacokinetic profiles of the analytes in normal and cholestatic liver injury (CLI) rats after oral administration of ZZDHD. Results showed that there were remarkable differences in pharmacokinetic properties of the analytes between normal and CLI group.