EMAIL THIS PAGE TO A FRIEND
Nature methods
Bone marrow-on-a-chip replicates hematopoietic niche physiology in vitro.
PMID 24793454
Abstract
Current in vitro hematopoiesis models fail to demonstrate the cellular diversity and complex functions of living bone marrow; hence, most translational studies relevant to the hematologic system are conducted in live animals. Here we describe a method for fabricating 'bone marrow-on-a-chip' that permits culture of living marrow with a functional hematopoietic niche in vitro by first engineering new bone in vivo, removing it whole and perfusing it with culture medium in a microfluidic device. The engineered bone marrow (eBM) retains hematopoietic stem and progenitor cells in normal in vivo-like proportions for at least 1 week in culture. eBM models organ-level marrow toxicity responses and protective effects of radiation countermeasure drugs, whereas conventional bone marrow culture methods do not. This biomimetic microdevice offers a new approach for analysis of drug responses and toxicities in bone marrow as well as for study of hematopoiesis and hematologic diseases in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1412069 ANTI-CXCL12 antibody produced in mouse, clone 2E2, purified immunoglobulin, buffered aqueous solution
SAB1412070
ANTI-CXCL12 antibody produced in mouse, clone 4D8, purified immunoglobulin, buffered aqueous solution
SAB1412168
ANTI-CXCL12 antibody produced in mouse, clone 1B2, purified immunoglobulin, buffered aqueous solution
SAB1412169
ANTI-CXCL12 antibody produced in mouse, clone 2A1, purified immunoglobulin, buffered aqueous solution
SAB1412233
ANTI-CXCL12 antibody produced in mouse, clone 2B1, purified immunoglobulin, buffered aqueous solution
SAB3501069
Anti-CXCL12 antibody produced in rabbit, affinity isolated antibody
GW21075 Anti-CXCR4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C3116
Anti-CXCR4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500383
Anti-CXCR4 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA030899
Anti-LEPR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2500591
Anti-Leptin Receptor antibody produced in goat, affinity isolated antibody, buffered aqueous solution
L9536 Anti-Leptin Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
HPA007007
Anti-NES antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA026111
Anti-NES antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104575
Anti-NES antibody produced in rabbit, affinity isolated antibody
N5413
Anti-Nestin antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
B5283 Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
R2655
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
WH0006387M1
Monoclonal Anti-CXCL12 antibody produced in mouse, clone 1E5, purified immunoglobulin, buffered aqueous solution
SAB1404360
Monoclonal Anti-CXCL12 antibody produced in mouse, clone 1F10, purified immunoglobulin, buffered aqueous solution
C6598 Monoclonal Anti-CXCR4 (Fusin) antibody produced in mouse, clone 44716, purified immunoglobulin, lyophilized powder
WH0007852M2
Monoclonal Anti-CXCR4 antibody produced in mouse, clone 1F8, ascites fluid
SAB1404560
Monoclonal Anti-CXCR4, (N-terminal) antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
SAB1402837
Monoclonal Anti-LEPR antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
AMAB90556
Monoclonal Anti-NES antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0197, purified immunoglobulin, buffered aqueous glycerol solution
R1008 Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
A2556 Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution
R3155 Monoclonal Anti-Rabbit IgG, Native−Peroxidase antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture