EMAIL THIS PAGE TO A FRIEND
Molecular neurobiology
Endoplasmic reticulum protein 29 protects cortical neurons from apoptosis and promoting corticospinal tract regeneration to improve neural behavior via caspase and Erk signal in rats with spinal cord transection.
PMID 24794144
Abstract
Following spinal cord injury (SCI), limit spontaneous functional recovery often emerged. However, the neuronal mechanisms associated with this phenomenon still remains obscure. By using proteomics analysis, endoplasmic reticulum protein 29 (ERp29) was discovered to increase in the motor cortexes of spinal cord transection (SCT) rats for 28 days post-operation (dpo) compared with in 14dpo. Then, the change in the expression of ERp29 was confirmed by using reverse transcription polymerase chain reaction (RT-PCR) and Western blot. To determine the role of ERp29 in the recovery of locomotor functions following SCT, lentiviral vectors were used to up- and downregulate the expression level of ERp29. Here, we found that cortical neurons in vitro with high level of ERp29 expression exhibited a significant proliferation, characterized by smaller size of soma and more extensive axon outgrowth, compared with neurons used as control, while ERp29 silence got the opposite results. In vivo, Lentivirus was inject into the cerebral cortex following SCT at thoracic level 10, which resulted in an increase number of neuronal nuclei(NeuN)-positive cells and less apoptotic cells. Moreover, increased PKC-γ immunoreactivity density was also found in the spinal cord T9 level compared with control rats. This was associated with a great functional improvement, indicated by Basso, Beattie, Bresnahan (BBB) locomotor rating scale. Lastly, we verified that ERp29 acts as a regulator by regulating a group of genes related with cell survival and apoptosis, involving in caspase and Erk, but not PI3K. Our findings showed that ERp29 can improve locomotor function by promoting neuronal survival and axonal regeneration in SCT rats via caspase and Erk signal pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA011868
Anti-DKK3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700747
Anti-DKK3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701251
Anti-DKK3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1408982
Anti-EREG antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4300344
Anti-ESR1 (Ab-104) antibody produced in rabbit, affinity isolated antibody
SAB4300345
Anti-ESR1 (Ab-106) antibody produced in rabbit, affinity isolated antibody
SAB4300346
Anti-ESR1 (Ab-118) antibody produced in rabbit, affinity isolated antibody
SAB4300347
Anti-ESR1 (Ab-167) antibody produced in rabbit, affinity isolated antibody
SAB2100711
Anti-ESR1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100712
Anti-ESR1 (ab2) antibody produced in rabbit, affinity isolated antibody
GW21182 Anti-ESR1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA000449
Anti-ESR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000450
Anti-ESR1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104971
Anti-ESR1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
E1396 Anti-Estrogen Receptor (154-171) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
E0521 Anti-Estrogen Receptor (18-32) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
P0050
Anti-Peroxiredoxin 6 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
AV48268
Anti-PRDX6 (AB1) antibody produced in rabbit, affinity isolated antibody
AV48267
Anti-PRDX6 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA006983
Anti-PRDX6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA030790
Anti-RBFOX3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA018265
Anti-SYF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500175
Anti-SYNGR1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA029673
Anti-SYNGR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5300114
Monoclonal Anti-DKK3 antibody produced in mouse, clone 4G7, ascites fluid
SAB1401128
Monoclonal Anti-EREG antibody produced in mouse, clone 1E6, purified immunoglobulin, buffered aqueous solution
AMAB90867
Monoclonal Anti-ESR1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1196, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402766
Monoclonal Anti-ESR1 antibody produced in mouse, clone 2F8, purified immunoglobulin, buffered aqueous solution
SAB5300303
Monoclonal Anti-ESR1 antibody produced in mouse, clone 2B7, purified immunoglobulin, buffered aqueous solution
SAB5300304
Monoclonal Anti-ESR1 antibody produced in mouse, clone 5D4B1, ascites fluid