EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Ultra-high performance liquid chromatography-quadrupole time-of-flight mass spectrometry to identify contaminants in water: an insight on environmental forensics.
PMID 24794942
Abstract
Ultra-high pressure liquid chromatography-quadrupole time-of-flight mass spectrometry (UHPLC-QqTOF-MS) acquiring full scan MS data for quantification, and automatic data dependent information product ion spectra (IDA-MS/MS) without any predefinition of the ions by the user was checked for identifying organic contaminants in water samples. The use of a database with more than 2000 compounds achieved high confidence results for a wide number of contaminants based upon retention time, accurate mass, isotopic pattern and MS/MS library searching. More than 20 contaminants, mostly pharmaceuticals, but also mycotoxins and polyphenols were unambiguously identified. Furthermore, the combination of statistical data analysis using principal component analysis (PCA) followed by empirical formula calculation, on-line database searching and MS/MS fragment ion interpretation achieves not only the successful detection of unknown contaminants but also the selection of those relevant to different types of waters. Unknown compounds, such as C₂₀H₃₄O₃, were identified in waste water showing the prospects of this technique. A group of 42 currently used pesticides were selected as target compounds to evaluate the quantitative possibilities. Mean recoveries and percentage relative standard deviation (RSD) were 48-79% (4-20% RSD). The limit of detections ranged from 0.02 to 2 ng L(-1), with a validated limit of quantification of 2 ng L(-1) for water after solid-phase (SPE) isolation and concentration. The quantitative data obtained using UHPLC-QqTOF-MS were compared with those obtained using conventional LC-MS/MS with a triple quadrupole (QqQ).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
516961
Ammonium formate, ≥99.995% trace metals basis
CH5NO2
09735
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
CH5NO2
70221
Ammonium formate, for mass spectrometry, ≥99.0%
CH5NO2
78314
Ammonium formate solution, BioUltra, 10 M in H2O
CH5NO2
714690
Ammonium formate solution, 10 mM in H2O, for HPLC
CH5NO2
66742
Dichloromethane, Selectophore, ≥99.5%
CH2Cl2
02575
Dichloromethane, analytical standard
CH2Cl2
45449
Dimethoate, PESTANAL®, analytical standard
C5H12NO3PS2
45477
Ethion, PESTANAL®, analytical standard
C9H22O4P2S4
N11964
Fenthion, analytical standard
C10H15O3PS2
36552
Fenthion, PESTANAL®, analytical standard
C10H15O3PS2
PS655
Fenthion, analytical standard
C10H15O3PS2
33365
Hexythiazox, PESTANAL®, analytical standard
C17H21ClN2O2S
PS1079
Hexythiazox, analytical standard
C17H21ClN2O2S
37894
Imidacloprid, PESTANAL®, analytical standard
C9H10ClN5O2
N12279
Isoproturon, analytical standard
C12H18N2O
36137
Isoproturon, PESTANAL®, analytical standard
C12H18N2O
36143
Malathion, PESTANAL®, analytical standard
C10H19O6PS2
91481
Malathion, certified reference material, TraceCERT®
C10H19O6PS2
1374408
Malathion, United States Pharmacopeia (USP) Reference Standard
C10H19O6PS2
SCM-039
Malathion, vial of 1 g, analytical standard
C10H19O6PS2
M0120000
Malathion, European Pharmacopoeia (EP) Reference Standard
C10H19O6PS2
36152
Methiocarb, PESTANAL®, analytical standard
C11H15NO2S
M1550000
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
CH2Cl2
PHR1557
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
CH2Cl2
N12478
Metolachlor, analytical standard
C15H22ClNO2
36163
Metolachlor, PESTANAL®, analytical standard
C15H22ClNO2
36171
Molinate, PESTANAL®, analytical standard
C9H17NOS
N12726
Omethoate, analytical standard
C5H12NO4PS
36181
Omethoate, PESTANAL®, analytical standard
C5H12NO4PS