EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
KIR/HLA interactions negatively affect rituximab- but not GA101 (obinutuzumab)-induced antibody-dependent cellular cytotoxicity.
PMID 24795454
Abstract
Ab-dependent cellular cytotoxicity (ADCC) mediated by NK cells is regulated by inhibitory killer cell Ig-like receptors (KIRs), which interact with target cell HLA class I. We analyzed how KIR/HLA interactions influence ADCC induced by rituximab and by GA101, a novel type II CD20 Ab glycoengineered for increased FcgRIII binding and ADCC capacity. We found that KIR/HLA interactions strongly and selectively inhibit rituximab-induced in vitro ADCC toward target cells expressing cognate HLA KIR ligands. NK cells of donors carrying all three ligands to inhibitory KIR showed weak activation and target cell depletion capacity when incubated with rituximab and KIR-ligand matched target B cells. In contrast, NK cells from individuals missing one or more KIR ligands activated more strongly and depleted KIR ligand-matched target B cells more efficiently in the presence of rituximab. NK cells expressing a KIR for which the ligand was absent were the main effectors of ADCC in these donors. Notably, the influence of KIR/HLA interactions on NK cell activation was synergistic with the effect of the V158F FCGR3A single nucleotide polymorphism. In contrast, GA101 induced activation of NK cells irrespective of inhibitory KIR expression, and efficiency of target cell depletion was not negatively affected by KIR/HLA interactions. These data show that modification of the Fc fragment to enhance ADCC can be an effective strategy to augment the efficacy of therapeutic mAbs by recruiting NK cells irrespective of their inhibitory KIR expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22783 Anti-CD14 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500215
Anti-CD14 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405579
Anti-CD14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001887
Anti-CD14 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002127
Anti-CD14 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW23003 Anti-CD19 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500285
Anti-LAMP-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
L1418
Anti-LAMP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA014750
Anti-LAMP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700418 Monoclonal Anti-CD107a-PE antibody produced in mouse, clone H4A3, purified immunoglobulin, buffered aqueous solution
AMAB90897
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1637, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90898
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1638, purified immunoglobulin, buffered aqueous glycerol solution
C7673
Monoclonal Anti-CD14 antibody produced in mouse, clone UCHM-1, purified immunoglobulin, buffered aqueous solution
SAB4700097
Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-15, purified immunoglobulin, buffered aqueous solution
SAB4700103 Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-18, purified immunoglobulin, buffered aqueous solution
SAB4700669 Monoclonal Anti-CD14 antibody produced in mouse, clone B-A8, purified immunoglobulin, buffered aqueous solution
WH0000930M1
Monoclonal Anti-CD19 antibody produced in mouse, clone 1G3, purified immunoglobulin, buffered aqueous solution
SAB1403645
Monoclonal Anti-CD19 antibody produced in mouse, clone 1C9, purified immunoglobulin, buffered aqueous solution
SAB4700108 Monoclonal Anti-CD19 antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
SAB4700109 Monoclonal Anti-CD19 antibody produced in mouse, clone LT19, purified immunoglobulin, buffered aqueous solution