EMAIL THIS PAGE TO A FRIEND
Oncotarget
Elevated Orai1 expression mediates tumor-promoting intracellular Ca2+ oscillations in human esophageal squamous cell carcinoma.
PMID 24797725
Abstract
Effective treatment as well as prognostic biomarker for malignant esophageal squamous cell carcinoma (ESCC) is urgently needed. The present study was aimed at identifying oncogenic genes involving dysregulated intracellular Ca2+ signaling, which is known to function importantly in cellular proliferation and migration. Tumors from patients with ESCC were found to display elevated expression of Orai1, a store-operated Ca2+ entry (SOCE) channel, and the high expression of Orai1 was associated with poor overall and recurrence-free survival. In contrast to the quiescent nature of non-tumorigenic epithelial cells, human ESCC cells exhibited strikingly hyperactive in intracellular Ca2+ oscillations, which were sensitive to treatments with Orai1 channel blockers and to orai1 silencing. Moreover, pharmacologic inhibition of Orai1 activity or reduction of Orai1 expression suppressed proliferation and migration of ESCC in vitro and slowed tumor formation and growth in in vivo xenografted mice. Combined, these findings provide the first evidence to imply Orai1 as a novel biomarker for ESCC prognostic stratification and also highlight Orai1-mediated Ca2+ signaling pathway as a potential target for treatment of this deadly disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CBL202
Anti-Vimentin Antibody, clone VIM 3B4, 50 µg, This Anti-Vimentin Antibody, clone VIM 3B4 is validated for use in ELISA, IF, IH(P), WB for the detection of Vimentin.
AV30161
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100360
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100378
Anti-CDC2 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100379
Anti-CDC2 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2103891
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
SAB4500051
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
SAB4503707
Anti-CDC2 antibody produced in rabbit, affinity isolated antibody
HPA003387
Anti-CDK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100391
Anti-CDKN1B antibody produced in rabbit, affinity isolated antibody
C8831
Anti-Cyclin B1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503501
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB4503502
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
AV50117
Anti-ORAI1 (AB1) antibody produced in rabbit, affinity isolated antibody
O8264
Anti-ORAI1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA016583
Anti-ORAI1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500412
Anti-ORAI1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300461
Anti-RAC1 (Ab-71) antibody produced in rabbit, affinity isolated antibody
SAB2106168
Anti-RAC1 antibody produced in rabbit, affinity isolated antibody
SAB4502560
Anti-RAC1 antibody produced in rabbit, affinity isolated antibody
HPA051979
Anti-TMEM132A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406585
Anti-VIM antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
WH0000891M1
Monoclonal Anti-CCNB1 antibody produced in mouse, clone 1C8, purified immunoglobulin, buffered aqueous solution
SAB5300329 Monoclonal Anti-CCNB1 antibody produced in mouse, clone 1B10, ascites fluid
SAB5300330
Monoclonal Anti-CCNB1 antibody produced in mouse, clone 5G6, ascites fluid
WH0006786M1
Monoclonal Anti-STIM1 antibody produced in mouse, clone 5A2, purified immunoglobulin, buffered aqueous solution
WH0007431M1
Monoclonal Anti-VIM antibody produced in mouse, clone 3E9, purified immunoglobulin, buffered aqueous solution
SAB4700524 Monoclonal Anti-VIM antibody produced in mouse, clone VI-RE/1, purified immunoglobulin, buffered aqueous solution
SAB4100612
Monoclonal Anti-VIM antibody produced in mouse, clone VIM-A604, culture supernatant
AMAB90516
Monoclonal Anti-VIM antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0157, purified immunoglobulin, buffered aqueous glycerol solution