EMAIL THIS PAGE TO A FRIEND
Oral diseases
The effect of five probiotic lactobacilli strains on the growth and biofilm formation of Streptococcus mutans.
PMID 24806217
Abstract
To compare the effects of five probiotic lactobacilli strains on the growth and biofilm formation of Streptococcus mutans (MS). Five probiotic lactobacilli bacteria (LB), Lactobacillus casei Shirota, Lactobacillus casei LC01, Lactobacillus plantarum ST-III, Lactobacillus paracasei Lpc-37, and Lactobacillus rhamnosus HN001, were used as test strains effecting on the Streptococci strain S. mutans UA159 in this study. The effect of LB strains and their supernatants on the viability of the MS was evaluated. Then, the effect of LB strains on the growth of MS biofilm formation was observed by fluorescence microscope. All of the LB strains inhibited the growth of MS at concentrations of 1 × 10(8) and 3 × 10(8) CFU ml(-1) (P < 0.05). Untreated (without pH adjustment and ultrafiltration) LB supernatants from all of the LB strains inhibited the growth of MS (P < 0.05) as well. After pH adjustment and ultrafiltration (treated), only supernatants from L. casei Shirota and L. rhamnosus HN001 inhibited the growth of MS (P < 0.05). MS biofilm formation was also inhibited by all untreated supernatants and by the treated supernatants of L. casei Shirota and L. rhamnosus HN001 (P < 0.05). All five probiotic lactobacilli strains inhibited the growth and biofilm formation of MS, likely through the production of an acid environment, bacteriocin-like poly peptides, or both, and the effects on MS were dependent on the LB strains used.