EMAIL THIS PAGE TO A FRIEND
International journal of urology : official journal of the Japanese Urological Association
Muscarinic receptors mediate cold stress-induced detrusor overactivity in type 2 diabetes mellitus rats.
PMID 24807830
Abstract
This study determined if muscarinic receptors could mediate the cold stress-induced detrusor overactivity induced in type 2 diabetes mellitus rats. Ten-week-old female Goto-Kakizaki diabetic rats (n = 12) and Wister Kyoto non-diabetic rats (n = 12) were maintained on a high-fat diet for 4 weeks. Cystometric investigations of the unanesthetized rats were carried out at room temperature (27 ± 2°C) for 20 min. They were intravenously administered imidafenacin (0.3 mg/kg, n = 6) or vehicle (n = 6). After 5 min, the rats were transferred to a low temperature (4 ± 2°C) for 40 min where the cystometry was continued. The rats were then returned to room temperature for the final cystometric measurements. Afterwards, expressions of bladder muscarinic receptor M3 and M2 messenger ribonucleic acids and proteins were assessed by reverse transcription polymerase chain reaction and immunohistochemistry. In non-diabetic Wister Kyoto rats, imidafenacin did not reduce cold stress-induced detrusor overactivity. In diabetic Goto-Kakizaki rats, just after transfer to a low temperature, the cold stress-induced detrusor overactivity in imidafenacin-treated rats was reduced compared with vehicle-treated rats. Within the urinary bladders, the ratio of M3 to M2 receptor messenger ribonucleic acid in the diabetic Goto-Kakizaki rats was significantly higher than that of the non-diabetic Wister Kyoto rats. The proportion of muscarinic M3 receptor-positive area within the detrusor in diabetic Goto-Kakizaki rats was also significantly higher than that in non-diabetic Wister Kyoto rats. Imidafenacin partially inhibits cold stress-induced detrusor overactivity in diabetic Goto-Kakizaki rats. In this animal model, muscarinic M3 receptors partially mediate cold stress-induced detrusor overactivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2502094
Anti-CHRM2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405619
Anti-CHRM2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA029795
Anti-CHRM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500245
Anti-CHRM2 antibody produced in rabbit, affinity isolated antibody
SAB1405620
Anti-CHRM3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA024106
Anti-CHRM3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
M9693 Anti-Muscarinic Acetylcholine Receptor M2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
M9568 Anti-Muscarinic Acetylcholine Receptor M3 antibody produced in rabbit, affinity isolated antibody
HPA018407
Anti-UPK3A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA018415
Anti-UPK3A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
WH0001129M1
Monoclonal Anti-CHRM2 antibody produced in mouse, clone 4B5, purified immunoglobulin, buffered aqueous solution