EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Protein phosphatase 6 controls BCR-induced apoptosis of WEHI-231 cells by regulating ubiquitination of Bcl-xL.
PMID 24808369
Abstract
Crosslinking BCR in the immature B cell line WEHI-231 causes apoptosis. We found that Bcl-xL was degraded by polyubiquitination upon BCR crosslinking and in this study explored the mechanism that controls the degradation of Bcl-xL. Ser(62) of Bcl-xL was phosphorylated by JNK to trigger polyubiquitination, and this was opposed by serine/threonine protein phosphatase 6 (PP6) that physically associated with Bcl-xL. We show BCR crosslinking decreased PP6 activity to allow Ser(62) phosphorylation of Bcl-xL. CD40 crosslinking rescues BCR-induced apoptosis, and we found PP6 associated with CD40 and PP6 activation in response to CD40. Our data suggest that PP6 activity is regulated to control apoptosis by modulating Ser(62) phosphorylation of Bcl-xL, which results in its polyubiquitination and degradation.