EMAIL THIS PAGE TO A FRIEND
CNS neuroscience & therapeutics
BASI, a potent small molecular inhibitor, inhibits glioblastoma progression by targeting microRNA-mediated β-catenin signaling.
PMID 24810017
Abstract
The nuclear localization of β-catenin, a mediator of canonical Wnt signaling, has been indicated in a variety of cancers and is frequently related to tumor progression and metastasis. Therefore, targeting β-catenin is an attractive therapeutic strategy for cancers. Herein, we identified a natural, small molecule inhibitor of β-catenin signaling, BASI, and evaluated its therapeutic efficacy both in vitro and in orthotopic mouse models of glioma. BASI significantly suppressed proliferation and invasion and induced apoptosis in glioblastoma cells and resulted in the remarkable attenuation of orthotopic tumor growth in vivo. Furthermore, we found that BASI altered the expression of several microRNAs, which mediated the posttranscriptional silencing of β-catenin expression either directly or indirectly through a von Hippel-Lindau (VHL)-mediated β-catenin degradation pattern. Taken together, our findings offer preclinical validation of BASI as a promising new type of β-catenin inhibitor with a mechanism of inhibition that has broad potential for the improved treatment of glioblastoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500454
Anti-CBP (Acetyl-Lys1535) antibody produced in rabbit, affinity isolated antibody
SAB4500455
Anti-CBP antibody produced in rabbit, affinity isolated antibody
SAB2100480
Anti-CREBBP antibody produced in rabbit, affinity isolated antibody
SAB4300485
Anti-EIF4E (Ab-209) antibody produced in rabbit, affinity isolated antibody
SAB1405749
Anti-EIF4E antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100666
Anti-EIF4E antibody produced in rabbit, affinity isolated antibody
SAB2107353
Anti-EIF4E antibody produced in rabbit, affinity isolated antibody
PRS2479
Anti-MCG10 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001632
Anti-PAG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107815
Anti-PAG1 antibody produced in rabbit, affinity isolated antibody
AV40568
Anti-PCBP2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV40567
Anti-PCBP2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA038356
Anti-PCBP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV40944
Anti-PCBP4 (AB1) antibody produced in rabbit, affinity isolated antibody
AV40943
Anti-PCBP4 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2500758
Anti-PCBP4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2900371
Anti-PCBP4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501099
Anti-VHL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406582
Anti-VHL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4200434
Anti-VHL antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4503075
Anti-VHL antibody produced in rabbit, affinity isolated antibody
WH0001387M1
Monoclonal Anti-CREBBP antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
SAB1403694
Monoclonal Anti-CREBBP antibody produced in mouse, clone 2B6, purified immunoglobulin, buffered aqueous solution
SAB5300107 Monoclonal Anti-EIF4E antibody produced in mouse, clone 5D11, ascites fluid
WH0005094M7
Monoclonal Anti-PCBP2 antibody produced in mouse, clone 5F12, purified immunoglobulin, buffered aqueous solution
SAB1404172
Monoclonal Anti-PCBP2 antibody produced in mouse, clone 3A1, purified immunoglobulin, buffered aqueous solution
WH0007428M1
Monoclonal Anti-VHL antibody produced in mouse, clone 1G12, purified immunoglobulin, buffered aqueous solution
PLA0097
Rabbit anti-CBP Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.