EMAIL THIS PAGE TO A FRIEND
Oncotarget
Long noncoding RNA ANRIL indicates a poor prognosis of gastric cancer and promotes tumor growth by epigenetically silencing of miR-99a/miR-449a.
PMID 24810364
Abstract
Long noncoding RNAs are involved in diseases including cancer. Here, we reported that ANRIL (CDKN2B-AS1), a 3.8-kb long noncoding RNA, recruiting and binding to PRC2, was generally upregulated in human gastric cancer (GC) tissues. In a cohort of 120 GC patients, the higher expression of ANRIL was significantly correlated with a higher TNM stage (P=0.041) and tumor size (P=0.001). Multivariate analyses revealed that ANRIL expression served as an independent predictor for overall survival (P=0.036). Further experiments revealed that ANRIL knockdown significantly repressed the proliferation both in vitro and in vivo. We also showed that E2F1 could induce ANRIL and ANRIL-mediated growth promotion is in part due to epigenetic repression of miR-99a/miR-449a in Trans (controlling the targets--mTOR and CDK6/E2F1 pathway) by binding to PRC2, thus forming a positive feedback loop, continuing to promote GC cell proliferation. To our knowledge, this is the first report showed that the role of ANRIL in the progression of GC and ANRIL could crosstalk with microRNAs in epigenetic level. Our results suggest that ANRIL, as a growth regulator, may serve as a candidate prognostic biomarker and target for new therapies in human gastric cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA008741
Anti-AGFG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21185 Anti-CDKN2A antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2502016
Anti-Cdkn2a antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501929
Anti-CDKN2A antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405602
Anti-CDKN2B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV03047 Anti-CDKN2B antibody produced in rabbit, affinity isolated antibody
SAB4500839
Anti-FAP-1 antibody produced in rabbit, affinity isolated antibody
SAB2101077
Anti-HRB antibody produced in rabbit, affinity isolated antibody
GW21943 Anti-MRPL28 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1407017
Anti-MRPL28 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA030594
Anti-MRPL28 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105816
Anti-MRPL28 antibody produced in rabbit, affinity isolated antibody
SAB4300187
Anti-phospho-RPS6 (pSer235) antibody produced in rabbit, affinity isolated antibody
GW21497A Anti-PTPN13 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW21551 Anti-PTPN13 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA006628
Anti-RBM14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV40635
Anti-RBM14 antibody produced in rabbit, affinity isolated antibody
AV37269
Anti-RBM14 antibody produced in rabbit, IgG fraction of antiserum
SAB1400734
Anti-RIP antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
GW21168 Anti-RIPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA015257
Anti-RIPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV02038
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody
SAB3500420
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA031526
Anti-RPAIN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
HPA023924
Anti-RPAIN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
AV39190
Anti-SUZ12 (AB2) antibody produced in rabbit, affinity isolated antibody
HPA057436
Anti-SUZ12 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3501103
Anti-SUZ12 antibody produced in rabbit, affinity isolated antibody
SAB5300498
Monoclonal Anti-CDKN2A antibody produced in mouse, clone 1E12E10, ascites fluid
SAB5300499
Monoclonal Anti-CDKN2A antibody produced in mouse, clone 5A8A4, ascites fluid