PloS one
A novel TetR-regulating peptide turns off rtTA-mediated activation of gene expression.
PMID 24810590
Abstract
Conditional regulation of gene expression is a powerful and indispensable method for analyzing gene function. The "Tet-On" system is a tool widely used for that purpose. Here, the transregulator rtTA mediates expression of a gene of interest after addition of the small molecule effector doxycycline. Although very effective in rapidly turning on gene expression, the system is hampered by the long half-life of doxycycline which makes shutting down gene expression rapidly very difficult to achieve. We isolated an rtTA-binding peptide by in vivo selection that acts as a doxycycline antagonist and leads to rapid and efficient shut down of rtTA-mediated reporter gene expression in a human cell line. This peptide represents the basis for novel effector molecules which complement the "Tet-system" by enabling the investigator to rapidly turn gene expression not just on at will, but now also off.