EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Human receptor activation by aroclor 1260, a polychlorinated biphenyl mixture.
PMID 24812009
Abstract
Polychlorinated biphenyls (PCBs) are persistent environmental toxicants, present in 100% ofxa0U.S. adults and dose-dependently associated with obesity and non-alcoholic fatty liver disease (NAFLD). PCBs are predicted to interact with receptors previously implicated in xenobiotic/energy metabolism and NAFLD. These receptors include the aryl hydrocarbon receptor (AhR), pregnane xenobiotic receptor (PXR), constitutive androstane receptor (CAR), peroxisome proliferator-activated receptors (PPARs), liver-X-receptor (LXRα), and farnesoid-X-receptor (FXR). This study evaluates Aroclor 1260, a PCB mixture with congener composition mimicking that of human adipose tissue, and selected congeners, as potential ligands for these receptors utilizing human hepatoma-derived (HepG2) and primate-derived (COS-1) cell lines, and primary human hepatocytes. Aroclor 1260 (20 μg/ml) activated AhR, and PCB 126, a minor component, was a potent inducer. Aroclor 1260 activated PXR in a simple concentration-dependent manner at concentrations ≥10 μg/ml. Among the congeners tested, PCBs 138, 149, 151, 174, 183, 187, and 196 activated PXR. Aroclor 1260 activated CAR2 and CAR3 variants at lower concentrations and antagonize CAR2 activation by the CAR agonist, CITCO, at higher concentrations (≥20 μg/ml). Additionally, Aroclor 1260 induced CYP2B6 in primary hepatocytes. At subtoxic doses, Aroclor 1260 did not activate LXR or FXR and had no effect on LXR- or FXR-dependent induction by the agonists T0901317 or GW4064, respectively. Aroclor 1260 (20 μg/ml) suppressed PPARα activation by the agonist nafenopin, although none of the congeners tested demonstrated significant inhibition. The results suggest that Aroclor 1260 is a human AhR, PXR and CAR3 agonist, a mixed agonist/antagonist for CAR2, and an antagonist for human PPARα.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B2209
Benz[a]anthracene, 99%
C18H12
48563
Benz[a]anthracene, analytical standard
C18H12
BCR271
Benz[a]anthracene, BCR® certified Reference Material
C18H12
75451
Benz[a]anthracene, certified reference material, TraceCERT®
C18H12
36956
Benz[a]anthracene solution, 100 ng/μL in cyclohexane, analytical standard
C18H12
48651
Benz[a]anthracene solution, certified reference material, 200 μg/mL in methylene chloride
C18H12
40070-U
Benz[a]anthracene solution, certified reference material, 1000 μg/mL in methanol
C18H12
242381
Benzoic acid, ACS reagent, ≥99.5%
C7H6O2
33045
Benzoic acid, for calorimetrical determination (approx. 26460 J/g)
C7H6O2
06185
Benzoic acid, Standard for quantitative NMR, TraceCERT®
C7H6O2
1055002
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O2
33047
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
C7H6O2
18102
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
C7H6O2
12356
Benzoic acid, tested according to Ph.Eur.
C7H6O2
PHR1050
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O2
12353
Benzoic acid, certified reference material for titrimetry, certified by BAM according to ISO 17025, ≥99.5%
C7H6O2
47849
Benzoic acid, analytical standard
C7H6O2
427608
Benzoic acid, purified by sublimation, ≥99%
C7H6O2
109479
Benzoic acid, ReagentPlus®, 99%
C7H6O2
W213101
Benzoic acid, ≥99.5%, FCC, FG
C7H6O2
W213128
Benzoic acid, natural, ≥99.5%, FCC, FG
C7H6O2
Y0001470
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
C7H6O2
491861
Benzoic acid-12C7, 99.9 atom % 12C
12C7H6O2
199524
Chlorophenol Red, indicator grade
C19H12Cl2O5S
C6240
CITCO, ≥98% (HPLC), solid
C19H12N3OCl3S
G5172
GW4064, ≥97% (HPLC)
C28H22Cl3NO4
76170
Melting point standard 121-123°C, analytical standard
C7H6O2
73983
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C7H6O2
46713
Rifampicin, VETRANAL, analytical standard
C43H58N4O12
R0700000
Rifampicin, European Pharmacopoeia (EP) Reference Standard
C43H58N4O12