EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Biodegradable nanoparticles composed of dendrigraft poly-L-lysine for gene delivery.
PMID 24813391
Abstract
We developed novel gene vectors composed of dendrigraft poly-L-lysine (DGL). The transgene expression efficiency of the pDNA/DGL complexes (DGL complexes) was markedly higher than that of the control pDNA/poly-L-lysine complex. However, the DGL complexes caused cytotoxicity and erythrocyte agglutination at high doses. Therefore, γ-polyglutamic acid (γ-PGA), which is a biodegradable anionic polymer, was added to the DGL complexes to decrease their toxicity. The resultant ternary complexes (DGL/γ-PGA complexes) were shown to be stable nanoparticles, and those with γ-PGA to pDNA charge ratios of >8 had anionic surface charges. The transgene expression efficiency of the DGL/γ-PGA complexes was similar to that of the DGL complexes; however, they exhibited lower cytotoxicity and did not induce erythrocyte agglutination at high doses. After being intravenously administered to mice, the DGL6 complex demonstrated high transfection efficiency in the liver, lungs, and spleen, whereas the DGL6/γ-PGA8 complex only displayed high transfection efficiency in the spleen. Future studies should examine the utility of DGL and DGL/γ-PGA complexes for clinical gene therapy.