EMAIL THIS PAGE TO A FRIEND
Acta tropica
Intranasal immunisation of the recombinant Toxoplasma gondii receptor for activated C kinase 1 partly protects mice against T. gondii infection.
PMID 24813415
Abstract
Nasal vaccination is an effective therapeutic regimen for preventing certain infectious diseases. The mucosal immune response is important for resistance to Toxoplasma gondii infection. In this study, we evaluated the immune responses elicited in BALB/c mice by nasal immunisation with recombinant T. gondii receptor for activated C kinase 1 (rTgRACK1) and their protective efficacy against T. gondii RH strain during both chronic and lethal infections. Nasal vaccination with rTgRACK1 increased the level of secretory IgA in nasal, intestinal and vesical washes, and the level of IFN-γ and IL-2 in intestinal washes, indicating that rTgRACK1 vaccination promotes mucosal immune responses. The mice immunised with rTgRACK1 also displayed increased levels of rTgRACK1-specific IgA, total IgG, IgG1 and in particular IgG2a in their blood sera, increased production of IFN-γ, IL-2 and IL-4 but not IL-10 from their isolated spleen cells, and enhanced splenocyte proliferation in vitro. rTgRACK1-vaccinated mice were effectively protected against infection with T. gondii RH strain, showing over 50% reduction of tachyzoite burdens in their liver and brain tissues during a chronic infection, and also a 45% increase in their survivals during a lethal challenge. These results indicate that rTgRACK1 might represent an intriguing immunogen for developing a mucosal vaccine against toxoplasmosis.