EMAIL THIS PAGE TO A FRIEND
Molecular therapy : the journal of the American Society of Gene Therapy
Systemic administration of a recombinant AAV1 vector encoding IGF-1 improves disease manifestations in SMA mice.
PMID 24814151
Abstract
Spinal muscular atrophy is a progressive motor neuron disease caused by a deficiency of survival motor neuron. In this study, we evaluated the efficacy of intravenous administration of a recombinant adeno-associated virus (AAV1) vector encoding human insulin-like growth factor-1 (IGF-1) in a severe mouse model of spinal muscular atrophy. Measurable quantities of human IGF-1 transcripts and protein were detected in the liver (up to 3 months postinjection) and in the serum indicating that IGF-1 was secreted from the liver into systemic circulation. Spinal muscular atrophy mice administered AAV1-IGF-1 on postnatal day 1 exhibited a lower extent of motor neuron degeneration, cardiac and muscle atrophy as well as a greater extent of innervation at the neuromuscular junctions compared to untreated controls at day 8 posttreatment. Importantly, treatment with AAV1-IGF-1 prolonged the animals' lifespan, increased their body weights and improved their motor coordination. Quantitative polymerase chain reaction and western blot analyses showed that AAV1-mediated expression of IGF-1 led to an increase in survival motor neuron transcript and protein levels in the spinal cord, brain, muscles, and heart. These data indicate that systemically delivered AAV1-IGF-1 can correct several of the biochemical and behavioral deficits in spinal muscular atrophy mice through increasing tissue levels of survival motor neuron.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS4235
Anti-Op18 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4237
Anti-Op18 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV40209
Anti-SMN1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA003482
Anti-SNRPB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406462
Anti-SNRPN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701198
Anti-SNRPN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300486
Anti-STMN1 (Ab-16) antibody produced in rabbit, affinity isolated antibody
SAB4300476
Anti-STMN1 (Ab-25) antibody produced in rabbit, affinity isolated antibody
SAB4300477
Anti-STMN1 (Ab-38) antibody produced in rabbit, affinity isolated antibody
SAB4300585
Anti-STMN1 (Ab-62) antibody produced in rabbit, affinity isolated antibody
SAB4501736
Anti-STMN1 antibody produced in rabbit, affinity isolated antibody
SAB2107336
Anti-STMN1 antibody produced in rabbit, affinity isolated antibody
S2944 Monoclonal Anti-SMN antibody produced in mouse, ~2 mg/mL, clone 2B1, purified immunoglobulin, buffered aqueous solution
SAB5300203 Monoclonal Anti-SMN1 antibody produced in mouse, clone 2F1, ascites fluid
SAB5300204 Monoclonal Anti-SMN1 antibody produced in mouse, clone 5H1, ascites fluid
WH0003925M1
Monoclonal Anti-STMN1 antibody produced in mouse, clone 3A9, ascites fluid, buffered aqueous solution
PLA0219
Rabbit anti-SMN1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.