EMAIL THIS PAGE TO A FRIEND
FEBS letters
Human Tudor staphylococcal nuclease (Tudor-SN) protein modulates the kinetics of AGTR1-3'UTR granule formation.
PMID 24815690
Abstract
Human Tudor staphylococcal nuclease (Tudor-SN) interacts with the G3BP protein and is recruited into stress granules (SGs), the main type of discrete RNA-containing cytoplasmic foci structure that is formed under stress conditions. Here, we further demonstrate that Tudor-SN binds and co-localizes with AGTR1-3'UTR (3'-untranslated region of angiotensin II receptor, type 1 mRNA) into SG. Tudor-SN plays an important role in the assembly of AGTR1-3'UTR granules. Moreover, endogenous Tudor-SN knockdown can decrease the recovery kinetics of AGTR1-3'UTR granules. Collectively, our data indicate that Tudor-SN modulates the kinetics of AGTR1-3'UTR granule formation, which provides an additional biological role of Tudor-SN in RNA metabolism during stress.