EMAIL THIS PAGE TO A FRIEND
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
The pro-oxidant buthionine sulfoximine (BSO) reduces tumor growth of implanted Lewis lung carcinoma in mice associated with increased protein carbonyl, tubulin abundance, and aminopeptidase activity.
PMID 24816945
Abstract
This study evaluated the effects of the pro-oxidant buthionine sulfoximine (BSO) and of the interaction between BSO and TETRAC, an antagonist of αvß3 integrin, on tumor development and aminopeptidase (AP) activity in a murine model of implanted Lewis's carcinoma. Male CBA-C57 mice were untreated (controls) or treated with BSO (222 mg/100 mL in drinking water), TETRAC (10 mg/kg/day, i.p.), or BSO + TETRAC. BSO for 28 days and TETRAC were given for the last 20 days. Mice were subcutaneously inoculated with 1 × 10(6) Lewis carcinoma 3LL cells into the dorsum. Study variables were tumor weight (TW); Hb, as index of tumor-mediated angiogenesis; vascular endothelial growth factor (VEGF) protein abundance; protein carbonyl content; α-tubulin abundance; and GluAp, AlaAp, and AspAp activities. BSO produced a major decrease in TW (203 ± 18 mg) with respect to controls (365 ± 26) and a reduction in Hb content. The TETRAC group also showed marked reductions in TW (129 ± 15) and Hb concentration associated with a reduced VEGF content. The BSO + TETRAC group showed a major TW reduction (125 ± 13); although, the difference with the TETRAC group was not significant. BSO treatment increased protein carbonyl and tubulin abundance in comparison to controls. The activity of all APs was increased in the three experimental groups and was strongly and negatively correlated with TW. In conclusion, administration of BSO reduced the TW, which inversely correlated with protein carbonyl content, suggesting a loss of microtubule polymerization. The finding of a negative correlation between TW and AP activity opens up new perspectives for the study of APs as tumor growth modulators.