EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Study of urinary steroid hormone disorders: difference between hepatocellular carcinoma in early stage and cirrhosis.
PMID 24817358
Abstract
Hepatocellular carcinoma (HCC) is the fifth most common cancer in the world. Discovery of novel biomarkers for early HCC from other liver diseases such as cirrhosis is of great clinical benefit. In this study, a novel steroid hormone metabolomic method based on liquid chromatography-mass spectrometry combined with logistic regression analysis was applied to study the steroid hormone disorders and to screen potential urinary steroid hormone biomarkers of early HCC. Thirty-six urinary steroid hormones were detected and quantified in healthy controls, cirrhotic patients, and early HCC patients. Heat map analysis and multivariate statistical analysis suggested severe disorders of steroid hormone network and holistically decreased urinary steroid hormone pattern in cirrhotic and early HCC patients. Logistic regression analysis reveals that a panel of two urinary steroid hormones (epitestosterone and allotetrahydrocortisol) displayed excellent diagnostic capability for distinguishing early HCC from cirrhosis with area under the curve (AUC) = 0.938 of receiver operating characteristic (ROC) analysis. These results help to overcome the disadvantage of lower sensitivity and specificity of alpha-fetoprotein for distinguishing early HCC from cirrhosis. Our work shows that steroid hormone metabolomics is a promising biomarker tool for biomarker study of early HCC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E3625
16-Epiestriol, ≥93% (HPLC)
C18H24O3
E3750
17-Epiestriol
C18H24O3
73532
2-Methoxyestrone, analytical standard
C19H24O3
D2004
4-(Dimethylamino)benzaldehyde, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
C9H11NO
33130
4-(Dimethylamino)benzaldehyde, puriss. p.a., Reag. Ph. Eur., ≥99% (perchloric acid titration)
C9H11NO
156477
4-(Dimethylamino)benzaldehyde, ACS reagent, 99%
C9H11NO
109762
4-(Dimethylamino)benzaldehyde, 98%
C9H11NO
02560
4-(Dimethylamino)benzaldehyde solution, 10 g/L in isopropanol, for TLC derivatization
C9H11NO
D139459
4-(Dimethylamino)benzoic acid, 98%
C9H11NO2
275654
4-(Dimethylamino)benzoic acid, 90%, technical grade
C9H11NO2
107700
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
C7H10N2
39405
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
C7H10N2
714844
4-(Dimethylamino)pyridine solution, 0.5 M in THF
C7H10N2
714720
4-(Dimethylamino)pyridine solution, 0.5 M in ethyl acetate
C7H10N2
700169
4-(Dimethylamino)pyridine, ChemDose tablets, Loading: 0.04 mmol tablet.
C7H10N2
P8754
5β-Pregnane-3α,17α,21-triol-20-one
C21H34O4
60983
Kovac’s reagent for indoles, for microbiology
C9H11NO
67309
Kovac’s reagent for indoles, for microbiology
C9H11NO
Y0001099
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
C7H10N2
1707894
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
C7H10N2