EMAIL THIS PAGE TO A FRIEND
Journal of cellular biochemistry
Altered signaling in the G1 phase deregulates chondrocyte growth in a mouse model with proteoglycan undersulfation.
PMID 24820054
Abstract
In several skeletal dysplasias defects in extracellular matrix molecules affect not only the structural and mechanical properties of cartilage, but also the complex network of signaling pathways involved in cell proliferation and differentiation. Sulfated proteoglycans, besides playing an important structural role in cartilage, are crucial in modulating the transport, diffusion, and interactions of growth factors with their specific targets, taking part in the regulation of signaling pathways involved in skeletal development and growth. In this work, we investigated by real time PCR and Western blots of the microdissected growth plate and by immunohistochemistry the molecular basis of reduced chondrocyte proliferation in the growth plate of the dtd mouse, a chondrodysplastic model with defective chondroitin sulfate proteoglycan sulfation of articular and growth plate cartilage. We detected activation of the Wnt pathway, leading to an increase in the non-phosphorylated form of nuclear β-catenin and subsequent up-regulation of cyclin D1 expression in the G1 phase of the cell cycle. β-Catenin was further stabilized by up-regulation of Smad3 expression through TGF-β pathway synergistic activation. We demonstrate that notwithstanding cyclin D1 expression increase, cell cycle progression is compromised in the G1 phase due to reduced phosphorylation of the pocket protein p130 leading to inhibition of transcription factors of the E2F family which are crucial for cell cycle progression and DNA replication. These data, together with altered Indian hedgehox signaling detected previously, explain at the molecular level the reduced chondrocyte proliferation rate of the dtd growth plate leading to reduced skeletal growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100362
Anti-CCND1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100363
Anti-CCND1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2100364
Anti-CCND1 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB1405510
Anti-CCND1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2106484
Anti-CCND1 antibody produced in rabbit, affinity isolated antibody
C5588 Anti-Cyclin D1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
N5162 Anti-Neurabin II antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1303872 ANTI-PLCL1 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA031849
Anti-PLCL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306820
ANTI-PPP1R9B(N-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500914
Anti-RAB3GAP1 antibody produced in rabbit, affinity isolated antibody
SAB2102983
Anti-RBL1 antibody produced in rabbit, affinity isolated antibody
HPA054962
Anti-RBL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA056525
Anti-RBL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502595
Anti-RBL1 antibody produced in rabbit, affinity isolated antibody
SAB2104003
Anti-RBL1, (N-terminal) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA019703
Anti-RBL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700017 Monoclonal Anti-CCND1 antibody produced in mouse, clone CD1.1, purified immunoglobulin, buffered aqueous solution
SAB5300525
Monoclonal Anti-CCND1 antibody produced in mouse, clone 3D8, ascites fluid
C7464
Monoclonal Anti-Cyclin D1 antibody produced in mouse, clone DCS-6, ascites fluid
P4360 Monoclonal Anti-p107 antibody produced in mouse, clone KAB6, purified immunoglobulin, buffered aqueous solution
P4485 Monoclonal Anti-p130 antibody produced in mouse, clone KAB40, purified immunoglobulin, buffered aqueous solution
WH0005933M1
Monoclonal Anti-RBL1 antibody produced in mouse, clone 1A5, purified immunoglobulin, buffered aqueous solution
SAB1409597
Monoclonal Anti-RBL2 antibody produced in mouse, clone 4D7, purified immunoglobulin, buffered aqueous solution
SAB1409598
Monoclonal Anti-RBL2 antibody produced in mouse, clone 2H7, purified immunoglobulin, buffered aqueous solution
SAB1409599 Monoclonal Anti-RBL2 antibody produced in mouse, clone 1F11, ascites fluid, solution