EMAIL THIS PAGE TO A FRIEND
Fertility and sterility
Estrogenic regulation of testicular expression of stem cell factor and c-kit: implications in germ cell survival and male fertility.
PMID 24825426
Abstract
To study the effect of estrogens regulating the testicular expression of stem cell factor (SCF) and c-kit. Experimental study. University research center. Male Wistar rats. Rat seminiferous tubules (SeT) cultured in the presence or absence of 17β-estradiol (E2). Expression of SCF and c-kit as well as apoptotic factors, FasL, FasR, Bcl-2, and Bax analyzed via quantitative reverse transcription-polymerase and Western blot; enzymatic activity of apoptosis effector caspase-3 assessed by colorimetric assay; proliferation index in SeT epithelium determined via fluorescent immunohistochemistry of nuclear proliferation marker Ki67. E2 (100xa0nM) induced a decrease in c-kit expression while increasing expression of SCF. Altered expression of the SCF/c-kit system relied on apoptosis of germ cells, as evidenced by the up-regulated expression of FasL/FasR, the increased ratio of proapoptotic/antiapoptotic proteins (Bax/Bcl-2), and the augmented activity of caspase-3. Decreased proliferation was also found in SeT in response to E2. A 100xa0nM dose of E2 unbalance the SCF/c-kit system, with a crucial impact on germ cell survival and thus male fertility. These findings contribute to our knowledge of the mechanisms underlying male idiopathic infertility associated with hyperestrogenism and open new perspectives on treatment targeting estrogen-signaling mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500343
Anti-Bax antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA027878
Anti-BAX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502549
Anti-BAX antibody produced in rabbit, affinity isolated antibody
PRS3335
Anti-Bcl-2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3337
Anti-Bcl-2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300339
Anti-BCL2 (Ab-56) antibody produced in rabbit, affinity isolated antibody
SAB4300340
Anti-BCL2 (Ab-70) antibody produced in rabbit, affinity isolated antibody
AV01005
Anti-BCL2 antibody produced in rabbit, affinity isolated antibody
HPA027444
Anti-FAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105218
Anti-FAS antibody produced in rabbit, affinity isolated antibody
SAB2101269
Anti-KITLG antibody produced in rabbit, affinity isolated antibody
SAB2101602
Anti-NKX3-1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101603
Anti-NKX3-1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB3500292
Anti-SCF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution