EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Identification of metabolites of N-(5-benzoyl-2-(4-(2-methoxyphenyl)piperazin-1-yl)thiazol-4-yl)pivalamide including CYP3A4-mediated C-demethylation in human liver microsomes with high-resolution/high-accuracy tandem mass.
PMID 24829290
Abstract
KRO-105714 [N-(5-benzoyl-2-(4-(2-methoxyphenyl)piperazin-1-yl)thiazol-4-yl)pivalamide] is a 2,4,5-trisubstituted 1,3-thiazole derivative that exerts anti-atopic dermatitis activity via robust suppression of the sphingosylphosphorylcholine receptor. This study used high-resolution/high-accuracy tandem mass spectroscopy (HRMS) and recombinant cDNA-expressed cytochrome P450 (P450) isoforms to identify the metabolic pathway and metabolites of KRO-105714 in human liver microsomes (HLMs) as therapeutic agents for inflammation. The incubation of KRO-105714 with pooled HLMs in the presence of NADPH generated four metabolites (M1-M4). The metabolites were identified using HRMS and confirmed using synthetic standards for M2 and M4. M1 and M2 were identified as monohydroxylated metabolites, and M3 and M4 were identified as O-demethyl KRO-105714 and C-demethyl KRO-105714, respectively. In the inhibition study with selective CYP3A4 inhibitors and incubation in recombinant cDNA-expressed P450 enzymes, all the metabolites of KRO-105714 were formed by CYP3A4 in HLMs. The CYP3A4-mediated formation of M4 from M2 was confirmed via incubation of M2 in HLMs. These results showed that the unusual C-demethylated metabolite M4 was generated from monohydroxyl metabolite M2 via a CYP3A4-mediated enzymatic reaction in HLMs.