EMAIL THIS PAGE TO A FRIEND
Canadian journal of physiology and pharmacology
Microinjection of l-glutamate into the nucleus ambiguus partially inhibits gastric motility through the NMDA receptor - nitric oxide pathway.
PMID 24830549
Abstract
We have previously reported that both l-glutamate (l-Glu) and nitric oxide (NO) modulate gastric motility in the nucleus ambiguus (NA). The aim of this study is to explore the potential correlation between the l-Glu and NO. A latex balloon connected to a pressure transducer was inserted into the pylorus through the fundus of anesthetized male Wistar rats to continuously record changes in gastric smooth muscle contractile curves. Pretreatment with the NO-synthase inhibitor N-nitro-l-arginine methylester (l-NAME) did not completely abolish the inhibitory effect of l-Glu on gastric motility, but intravenous injection of the ganglionic blocker hexamethonium bromide (Hb) did. By using a specific N-methyl-d-aspartic acid (NMDA) receptor antagonist, we blocked the inhibitory effect of the NO-donor sodium nitroprusside (SNP) on gastric motility. These results suggest that microinjections of l-Glu into the NA inhibits gastric motility by activating the cholinergic preganglionic neurons, partially through the NMDA receptor - NO pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0879
Hexamethonium bromide
C12H30Br2N2
52590
Hexamethonium bromide, ≥95.0% (AT)
C12H30Br2N2