EMAIL THIS PAGE TO A FRIEND
Breast cancer research and treatment
Novel genetic aberrations in breast phyllodes tumours: comparison between prognostically distinct groups.
PMID 24831776
Abstract
Phyllodes tumours of the breast are uncommon fibroepithelial neoplasms which pose management challenges due to difficulties in accurate prediction of clinical behaviour, as histological assessment has its limitations. Molecular studies have improved the understanding of these rare tumours but such findings are scant. We aimed to investigate genetic aberrations in phyllodes tumours stratified according to clinical behaviour, to identify potential genes contributing to disease progression. Twenty phyllodes tumours were separated into prognostically distinct categories depending on whether they had recurred/metastasized within the follow-up period. DNA extracted from FFPE materials was subjected to Affymetrix OncoScan™ FFPE Express molecular inversion probe microarray platform for analysis of copy number changes and mutational status. Results were cross validated with Sanger sequencing, FISH and immunohistochemistry. A higher number of chromosomal aberrations were observed in cases which recurred/metastasized, with median events of 19 compared to 3.5 in cases which did not recur/metastasize. High-level amplification and homozygous deletions were detected exclusively in the former group. Regions of high-level amplification included MDM4 (1q32.1), RAF1 (3p25), EGFR (7p12) and PDZD2 (5p13.3). EGFR amplification was confirmed on FISH and accompanied by intense EGFR immunostaining. Regions of homozygous deletion included CDKN2A (9p21) and MACROD2 (20p12.1). Homozygous deletion of 9p21 which involved CDKN2A was accompanied by loss of protein expression. No mutations were identified in all samples. These findings provide insights into identifying target genes and pathways exploited by phyllodes tumours, which would aid future development of individualised therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300352
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
SAB4300353
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
SAB4300510
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
SAB4300472
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAB4300602
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
SAB4300480
Anti-EGFR (Ab-1197) antibody produced in rabbit, affinity isolated antibody
SAB4300453
Anti-EGFR (Ab-678) antibody produced in rabbit, affinity isolated antibody
SAB4300454
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
SAB4300481
Anti-EGFR (Ab-869) antibody produced in rabbit, affinity isolated antibody
GW21146 Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405746
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001200
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA018530
Anti-EGFR antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500711
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500712
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500720
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
WH0001956M2 Monoclonal Anti-EGFR antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
AMAB90816
Monoclonal Anti-EGFR antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0815, purified immunoglobulin, buffered aqueous glycerol solution
E2760
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid
E3138
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid
E0532 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 102618, purified immunoglobulin, lyophilized powder
PLA0101
Rabbit anti-EGFR Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.