EMAIL THIS PAGE TO A FRIEND
Biology
Application of fluorescence two-dimensional difference in-gel electrophoresis as a proteomic biomarker discovery tool in muscular dystrophy research.
PMID 24833232
Abstract
In this article, we illustrate the application of difference in-gel electrophoresis for the proteomic analysis of dystrophic skeletal muscle. The mdx diaphragm was used as a tissue model of dystrophinopathy. Two-dimensional gel electrophoresis is a widely employed protein separation method in proteomic investigations. Although two-dimensional gels usually underestimate the cellular presence of very high molecular mass proteins, integral membrane proteins and low copy number proteins, this method is extremely powerful in the comprehensive analysis of contractile proteins, metabolic enzymes, structural proteins and molecular chaperones. This gives rise to two-dimensional gel electrophoretic separation as the method of choice for studying contractile tissues in health and disease. For comparative studies, fluorescence difference in-gel electrophoresis has been shown to provide an excellent biomarker discovery tool. Since aged diaphragm fibres from the mdx mouse model of Duchenne muscular dystrophy closely resemble the human pathology, we have carried out a mass spectrometry-based comparison of the naturally aged diaphragm versus the senescent dystrophic diaphragm. The proteomic comparison of wild type versus mdx diaphragm resulted in the identification of 84 altered protein species. Novel molecular insights into dystrophic changes suggest increased cellular stress, impaired calcium buffering, cytostructural alterations and disturbances of mitochondrial metabolism in dystrophin-deficient muscle tissue.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45983
Acetonitrile, analytical standard
C2H3N
PHR1551
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
00712
Acetonitrile solution, ~20% in H2O, for protein sequence analysis
C2H3N
A6390
β-D-Allose, rare aldohexose sugar
C6H12O6
62860
DL-Lysine, purum, ≥95.0% (NT)
C6H14N2O2
49897
Formate standard for IC, 1.000 g/L in H2O, analytical standard
CH2O2
27002
Formic acid, purum, ≥85%
CH2O2
06473
Formic acid solution, BioUltra, 1.0 M in H2O
CH2O2
1295607
Glycerin, United States Pharmacopeia (USP) Reference Standard
C3H8O3
PHR1020
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O3
49779
Glycerol, tested according to Ph.Eur., anhydrous
C3H8O3
77067
Glycerol, analytical standard
C3H8O3
12146
Glycerol 3 M solution, 3 M
C3H8O3 · xH2O
PHR1320
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
C2H3N
1601340
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
C2H3N
U0600000
Urea, European Pharmacopoeia (EP) Reference Standard
CH4N2O
1706698
Urea, United States Pharmacopeia (USP) Reference Standard
CH4N2O
56180
Urea, analytical standard
CH4N2O
51465
Urea, tested according to Ph.Eur.
CH4N2O
08582
Urea solution, for microbiology, 40% in H2O
CH4N2O