EMAIL THIS PAGE TO A FRIEND
Journal of applied physiology (Bethesda, Md. : 1985)
Activating cAMP/PKA signaling in skeletal muscle suppresses the ubiquitin-proteasome-dependent proteolysis: implications for sympathetic regulation.
PMID 24833777
Abstract
Although we have recently demonstrated that plasma catecholamines induce antiproteolytic effects on skeletal muscle (Graça FA, Gonçalves DAP, Silveira WA, Lira EC, Chaves VE, Zanon NM, Garófalo MAR, Kettelhut IC, Navegantes LCC. Am J Physiol Endocrinol Metab. 305: E1483-E1494, 2013), the role of the muscle sympathetic innervation and, more specifically, norepinephrine (NE) in regulating the ubiquitin (Ub)-proteasome system (UPS) remains unknown. Based on previous findings that chemical sympathectomy acutely reduces UPS activity, we hypothesized that muscle NE depletion induces adrenergic supersensitivity in rat skeletal muscles. We report that surgical sympathetic denervation (SDEN), a condition in which only muscle NE from both hindlimbs is depleted, transiently reduced the overall proteolysis and the UPS activity (∼25%) in both soleus and extensor digitorum longus muscles. This antiproteolytic response was accompanied by increased activity of adenylyl cyclase (112%), levels of cyclic adenosine monophosphate (cAMP; 191%), and the serine phosphorylation of cAMP response element-binding protein (32%). In extensor digitorum longus from normal rats, NE (10(-4) M) in vitro increased the levels of cAMP (115%) and the serine phosphorylation of both cAMP response element-binding protein (2.7-fold) and forkhead box class O1 transcription factor. Similar effects were observed in C2C12 cells incubated with forskolin (10 μM). In parallel, NE significantly reduced the basal UPS (21%) activity and the mRNA levels of atrophy-related Ub-ligases. Similar responses were observed in isolated muscles exposed to 6-BNZ-cAMP (500 μM), a specific PKA activator. The phosphorylation levels of Akt were not altered by SDEN, NE, forskolin or 6-BNZ-cAMP. Our results demonstrate that SDEN induces muscle adrenergic supersensitivity for cAMP leading to the suppression of UPS, and that the suppressive effects of NE on UPS activity and expression of Ub-ligases can be mediated by the activation of cAMP/PKA signaling, with the inhibition of forkhead box class O1 transcription factor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306343
ANTI-FKHR (CENTER D469) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
F2303 Anti-FKHR (FOXO1a) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500938
Anti-FKHR antibody produced in rabbit, affinity isolated antibody
SAB4500939
Anti-FKHR antibody produced in rabbit, affinity isolated antibody
SAB4300410
Anti-FOXO1 (Ab-256) antibody produced in rabbit, affinity isolated antibody
SAB4300428
Anti-FOXO1 (Ab-319) antibody produced in rabbit, affinity isolated antibody
AV100607
Anti-FOXO1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV32107
Anti-FOXO1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV32976
Anti-FOXO1 (AB3) antibody produced in rabbit, affinity isolated antibody
SAB2500417
Anti-FOXO1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001252
Anti-FOXO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500507
Anti-FOXO1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1411790
ANTI-FOXO1A antibody produced in mouse, clone 4B4, purified immunoglobulin, buffered aqueous solution
SAB1411812
ANTI-FOXO1A antibody produced in mouse, clone 3H5, purified immunoglobulin, buffered aqueous solution
SAB1411813
ANTI-FOXO1A antibody produced in mouse, clone 4A3, purified immunoglobulin, buffered aqueous solution
SAB1411814
ANTI-FOXO1A antibody produced in mouse, clone 4A10, purified immunoglobulin, buffered aqueous solution
SAB1411815
ANTI-FOXO1A antibody produced in mouse, clone 2F12, purified immunoglobulin, buffered aqueous solution
SAB1411857
ANTI-FOXO1A antibody produced in mouse, clone 1F11, purified immunoglobulin, buffered aqueous solution
SAB1411858
ANTI-FOXO1A antibody produced in mouse, clone 4A2, purified immunoglobulin, buffered aqueous solution
SAB1411859
ANTI-FOXO1A antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
SAB1411860
ANTI-FOXO1A antibody produced in mouse, clone 1A6, purified immunoglobulin, buffered aqueous solution
SAB1411861
ANTI-FOXO1A antibody produced in mouse, clone 1F6, purified immunoglobulin, buffered aqueous solution
AV38094
Anti-FOXO1A antibody produced in rabbit, IgG fraction of antiserum
SAB4500936
Anti-FOXO1A antibody produced in rabbit, affinity isolated antibody
SAB4500937
Anti-FOXO1A antibody produced in rabbit, affinity isolated antibody
C9102 Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504375
Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB1306301 ANTI-PHOSPHO-CREB(S133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300040
Anti-phospho-CREB1 (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB4504407
Anti-phospho-FKHR (pSer256) antibody produced in rabbit, affinity isolated antibody