EMAIL THIS PAGE TO A FRIEND
Respiratory medicine
Baseline KL-6 predicts increased risk for acute exacerbation of idiopathic pulmonary fibrosis.
PMID 24835074
Abstract
Acute exacerbation (AE) is a major cause of death in idiopathic pulmonary fibrosis (IPF). However, little is known about sensitive biomarkers for predicting AE. The aim of our study was to investigate the significance of KL-6 and CC-Chemokine Ligand 18 (CCL18) as predictors for AE of IPF. We prospectively collected a total of 77 patients with IPF. Serum levels of KL-6 and CCL18 were measured by ELISA. The correlation between baseline serum levels of the markers and the incidence of AE was evaluated. Thirteen (17%) patients experienced AE during follow-up. Baseline serum KL-6 levels were significantly higher in patients who developed AE than in patients with stable IPF (pxa0
Related Materials