EMAIL THIS PAGE TO A FRIEND
Journal of controlled release : official journal of the Controlled Release Society
Prolonged blood circulation and enhanced tumor accumulation of folate-targeted dendrimer-polymer hybrid nanoparticles.
PMID 24837188
Abstract
Nanoparticle (NP)-based drug delivery platforms have received a great deal of attention over the past two decades for their potential in targeted cancer therapies. Despite the promises, passive targeting approaches utilizing relatively larger NPs (typically 50-200nm in diameter) allow for passive tumor accumulation, but hinder efficient intratumoral penetration. Conversely, smaller, actively targeted NPs (<20nm in diameter) penetrate well into the tumor mass, but are limited by their rapid systemic elimination. To overcome these limitations, we have designed a multi-scale hybrid NP platform that loads smaller poly(amidoamine) (PAMAM) dendrimers (~5nm in diameter) into larger poly(ethylene glycol)-b-poly(D,L-lactide) (PEG-PLA) NPs (~70nm). A biodistribution study in healthy mice revealed that the hybrid NPs circulated longer than free dendrimers and were mostly cleared by macrophages in the liver and spleen, similar to the in vivo behavior of PEG-PLA NPs. When injected intravenously into the BALB/c athymic nude mice bearing folate receptor (FR)-overexpressing KB xenograft, the targeted hybrid NPs encapsulating folate (FA)-targeted dendrimers achieved longer plasma circulation than free dendrimers and higher tumor concentrations than both free dendrimers and the empty PEG-PLA NPs. These results suggest that the hybrid NPs successfully combine the in vivo advantages of dendrimers and polymeric NPs, demonstrating their potential as a new, modular platform for drug delivery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74595
(±)-Glycidol, analytical standard
C3H6O2
74440
(−)-Nopol, purum, ≥98.0% (sum of enantiomers, GC)
C11H18O
66742
Dichloromethane, Selectophore, ≥99.5%
CH2Cl2
02575
Dichloromethane, analytical standard
CH2Cl2
38911
Dimethyl ether, puriss., ≥99.9% (GC)
C2H6O
295299
Dimethyl ether, ≥99%
C2H6O
PHR1553
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO
F7876
Folic acid, ≥97%
C19H19N7O6
1286005
Folic acid, United States Pharmacopeia (USP) Reference Standard
C19H19N7O6
F8758
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
C19H19N7O6
F8798
Folic acid, meets USP testing specifications
C19H19N7O6
01769
Folic acid, tested according to Ph.Eur.
C19H19N7O6
F0300000
Folic acid, European Pharmacopoeia (EP) Reference Standard
C19H19N7O6
PHR1035
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C19H19N7O6
47866
Folic acid, analytical standard
C19H19N7O6
G5809
Glycidol, 96%
C3H6O2
Y0001048
Levodropropizine impurity C, European Pharmacopoeia (EP) Reference Standard
C3H6O2
M1550000
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
CH2Cl2
PHR1557
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
CH2Cl2
72438
N,N-Dimethylformamide, analytical standard
C3H7NO
283924
Rhodamine B isothiocyanate, mixed isomers
C29H30ClN3O3S
R1755
Rhodamine B isothiocyanate, BioReagent, mixed isomers, suitable for protein labeling
C29H30ClN3O3S