EMAIL THIS PAGE TO A FRIEND
Drug and alcohol dependence
Relaxin-3 mRNA levels in nucleus incertus correlate with alcohol and sucrose intake in rats.
PMID 24837581
Abstract
Chronic alcohol intake produces multiple neuroadaptive changes, including up- and down-regulation of neuropeptides and receptors. There are widespread projections of relaxin-3 containing neurons to, and abundant relaxin family peptide 3 receptor (RXFP3) expression within, brain regions involved in modulating alcohol intake. Recently we demonstrated the involvement of relaxin-3/RXFP3 signalling in alcohol-seeking in rats; therefore in this study we examined whether relaxin-3 and/or RXFP3 expression were altered by chronic alcohol intake in alcohol-preferring iP rats. Expression of relaxin-3 mRNA in the hindbrain nucleus incertus and RXFP3 radioligand binding levels in discrete forebrain regions were investigated following voluntary intake of alcohol or sucrose for 12 weeks, with a 2 day washout, using quantitative in situ hybridisation histochemistry and in vitro receptor autoradiography, respectively, in cohorts of adult, male iP rats. Levels of relaxin-3 mRNA in the hindbrain nucleus incertus were positively correlated with the level of intake of both alcohol (r(12)=0.59, p=0.03) and sucrose (r(7)=0.70, p=0.04) in iP rats. Dense binding of the RXFP3-selective radioligand, [(125)]-R3/I5, was detected in hypothalamic and extrahypothalamic sites, but no significant changes in the density of RXFP3 were observed in the brain regions quantified following chronic sucrose or ethanol intake. Our findings suggest high endogenous relaxin-3 expression may be associated with higher intake of rewarding substances, rather than its expression being regulated in response to their intake, consistent with an active role for the relaxin-3/RXFP3 system in modulating ingestive and alcohol-related behaviours.