EMAIL THIS PAGE TO A FRIEND
Immunologic research
Increased levels of anti-heat-shock protein 60 (anti-Hsp60) indicate endothelial dysfunction, atherosclerosis and cardiovascular diseases in patients with mixed connective tissue disease.
PMID 24838263
Abstract
Heat-shock protein 60 (Hsp60) has been shown to provoke inflammation, and anti-Hsp60 may facilitate the development of atherosclerosis. In this study, we have investigated 30 patients with mixed connective tissue disease (MCTD) and assessed anti-Hsp60 and their relationship to cardiovascular diseases (CVD). Out of 30 patients with MCTD, 15 had CVDs. Anti-Hsp60 antibody was determined by enzyme-linked immunosorbent assay. Since endothelial dysfunction and accelerated atherosclerosis are characteristic to MCTD, a wide array of MCTD-, endothelial dysfunction- and CVD-associated parameters was investigated: serum lipid levels, paraoxonase activity (PON1), rich nuclear ribonucleoprotein U1 (anti-U1RNP), anti-endothelial cell antibodies, anti-cardiolipin and anti-β2-glycoprotein I antibody isotypes (anti-CL and anti-β2GPI), endothelin-1 (ET-1) levels, also intima-media thickness (IMT), a quantitative indicator of atherosclerosis. In MCTD, anti-Hsp60 antibody levels were significantly higher than in healthy individuals (pxa0