EMAIL THIS PAGE TO A FRIEND
Digestive diseases and sciences
Specific expression of apolipoprotein A-IV in the follicle-associated epithelium of the small intestine.
PMID 24838500
Abstract
Peyer's patches (PPs), which are covered by specialized follicle-associated epithelium (FAE) including M cells, play a central role in immune induction in the gastrointestinal tract. This study is to investigate a new molecule to characterize PPs. We generated a monoclonal antibody (mAb 10-15-3-3) that specifically reacts to the epithelium of PPs and isolated lymphoid follicles. Target antigen was analyzed by immunoprecipitation and mass spectrometry. Localization and expression of target antigen were evaluated by immunofluorescence, in situ hybridization and real-time PCR. Immunoprecipitation and mass spectrometry revealed that mAb 10-15-3-3 recognized apolipoprotein A-IV (ApoA-IV), a well-known lipid transporter; this finding was confirmed by the specific reactivity of mAb 10-15-3-3 to cells transfected with the murine ApoA-IV gene. Immunofluorescence using mAb 10-15-3-3 showed intestinal localization of ApoA-IV, in which strong expression of the ApoA-IV protein occurred throughout the entire intestinal epithelium during developing period before weaning but was restricted to the FAE in adult mice. In support of these findings, in situ hybridization showed strong expression of the ApoA-IV gene throughout the entire intestinal epithelium during developing period before weaning, but this expression was restricted to the FAE predominantly and the tips of villi to a lesser extent in adult mice. Deficiency of ApoA-IV had no effect on the organogenesis of PP in mice. Our current results reveal ApoA-IV as a novel FAE-specific marker especially in the upper small intestine of adult mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4700041 Anti-CD3, low endotoxin antibody, Mouse monoclonal, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4300686
Anti-CD3E antibody produced in rabbit, affinity isolated antibody
D9026
Digoxigenin, analytical standard
C23H34O5
Y0000640
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
C23H34O5
44208
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
C6H14N2O2 · HCl
PHR1101
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14N2O2 · HCl
67448
L-Lysine monohydrochloride, certified reference material, TraceCERT®
C6H14N2O2 · HCl
L0900000
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C6H14N2O2 · HCl
C7048
Monoclonal Anti-CD3 antibody produced in mouse, clone UCHT-1, purified immunoglobulin, buffered aqueous solution
SAB4700040 Monoclonal Anti-CD3 antibody produced in mouse, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4700044 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-57, purified immunoglobulin, buffered aqueous solution
SAB4700045 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-92, purified immunoglobulin, buffered aqueous solution
SAB4700050 Monoclonal Anti-CD3 antibody produced in mouse, clone APA1/1, purified immunoglobulin, buffered aqueous solution
WH0000916M4
Monoclonal Anti-CD3E antibody produced in mouse, clone 4C1, purified immunoglobulin, buffered aqueous solution
AMAB90876
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1466, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90879
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1497, purified immunoglobulin, buffered aqueous glycerol solution