EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Human papillomavirus DNA load and p16INK4a expression predict for local control in patients with anal squamous cell carcinoma treated with chemoradiotherapy.
PMID 24839133
Abstract
As the detection rate of HPV-DNA in anal carcinoma commonly exceeds 90%, a comparison between sole HPV-positive and HPV-negative cancers with respect to treatment response following chemoradiotherapy (CRT) and long-term oncological outcome is challenging. Against this background, we aimed to assess HPV types and HPV DNA load in formalin-fixed paraffin-embedded tissue (FFPE) of 95 patients treated with standard CRT for anal cancer to correlate viral load (≤/> median) with local failure, distant metastases, cancer-specific (CSS) and overall survival (OS) rates. Various clinicopathologic parameters and the immunohistochemical marker p16(INK4a) were evaluated for any correlation with HPV16 DNA load and were included in uni- and multivariate analyses. The overall prevalence of HPV DNA was 95.8% with HPV16 monoinfection being the most commonly encountered HPV type (78.9%), followed by HPV16 and 31, 35, 39, 44, 58, 66 and 81 dual infection in 9 patients (9.5%). HPV16 DNA load was significantly associated with p16(INK4a) expression (p = 0.001). Patients with HPV16 DNA load ≤ median and low p16(INK4a) expression showed significantly worse local control (HPV16 DNA load: univariate p = 0.023, multivariate p = 0.042; p16(INK4a): univariate p = 0.021), and OS (HPV16 DNA load: univariate p = 0.02, multivariate p = 0.03). Moreover, a combined HPV16 DNA load and p16(INK4a) variable revealed a significant correlation to decreased local failure, and increased CSS and OS (p = 0.019, p = 0.04 and p = 0.031). In conclusion, these data indicate that HPV16 DNA load and p16(INK4a) expression are significant prognostic factors for local tumor control and overall survival of patients with anal SCC following CRT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000378 Mitomycin, European Pharmacopoeia (EP) Reference Standard
C15H18N4O5