EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Effect of processing conditions on the organosulfides of shallot (Allium cepa L. Aggregatum group).
PMID 24840922
Abstract
There is a growing account of the health benefits of H2S as an endogenous cell-signaling molecule. H2S from organic polysulfides, in particular, is increasingly gaining attention for their beneficial effects to cardiovascular health. Here, we studied shallot as a potential dietary source of organic polysulfides and examined the effects of processing conditions on its polysulfide profiles. Boiling, autoclaving, and freeze-drying were tested on whole and crushed shallot bulbs, analyzing their effect on the yield of organosulfides. Seventeen organosulfides, including disulfides, trisulfides, and cyclic polysulfides, were identified. Significant differences in the quantitative and qualitative profiles of organosulfides in the hydrodistilled and solvent extracted oils were observed. Freeze-drying retained the majority of the organosulfides, but the whole-autoclaved and whole-boiled shallots lost more than 95% of their organic polysulfides. Crushed-boiled and crushed-autoclaved shallot lost 76-80% of their organosulfides, likely due to the thermal sensitivity of these compounds. The organosulfide profiles are sensitive to the pH values of the processing media. In general, disulfides increased at basic pH (pH 9.0) while trisulfides and cyclic organosulfides are much higher at the acidic to neutral pH values (pH 3.0-5.0). Our results provide important information on the effects of processing conditions that are relevant for optimizing extraction of organosulfides from shallot for further studies evaluating their H2S-releasing activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W515507
2,5-Dimethylthiophene, ≥98%
C6H8S
D188603
2,5-Dimethylthiophene, 98.5%
C6H8S
220116
2-Hydroxybutyric acid sodium salt, 97%
C4H7NaO3
317691
Allyl disulfide, technical grade, 80%
C6H10S2
W202800
Allyl disulfide, ≥80%, FG
C6H10S2
38730 Citrate standard for IC, 1000 mg/L, analytical standard
C6H8O7
791725
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
C6H8O7
C2404
Citric acid, anhydrous, cell culture tested, plant cell culture tested
C6H8O7
C83155
Citric acid, 99%
C6H8O7
C0759
Citric acid, 99%
C6H8O7
94676
Citric acid, certified reference material, TraceCERT®
C6H8O7
46933
Citric acid, analytical standard
C6H8O7
1134368
Citric acid, United States Pharmacopeia (USP) Reference Standard
C6H8O7
27487
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
C6H8O7
A1202000
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
C6H8O7
W230618
Citric acid, ≥99.5%, FCC, FG
C6H8O7
PHR1071
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
C6H8O7
66742
Dichloromethane, Selectophore, ≥99.5%
CH2Cl2
02575
Dichloromethane, analytical standard
CH2Cl2
48538
Dichloromethane, analytical standard, stabilized
CH2Cl2
48600
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
CH2Cl2
40042
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
CH2Cl2
296082
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
C4H10O
91238
Diethyl ether, analytical standard
C4H10O
09-1833
Diethyl ether, for residue analysis, suitable for 5000 per JIS
C4H10O
09-1855
Diethyl ether
C4H10O
471569
Dimethyl disulfide, ≥99.0%
C2H6S2
320412
Dimethyl disulfide, 99%
C2H6S2
W353604
Dimethyl disulfide, ≥98%, FG
C2H6S2
68986
Dimethyl disulfide, analytical standard
C2H6S2