EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Biotransformation of glucoaurantio-obtusin towards aurantio-obtusin increases the toxicity of irinotecan through increased inhibition towards SN-38 glucuronidation.
PMID 24842785
Abstract
The present study aims to investigate the influence of irinotecan's toxicity by the biotransformation of glucoaurantio-obtusin to aurantio-obtusin. Intraperitoneal administration (i.p.) of 100 mg/kg aurantio-obtusin significantly increased the toxicity of irinotecan, but the i.p. administration of 100 mg/kg glucoaurantio-obtusin showed negligible influence towards irinotecan's toxicity. Furthermore, the mechanism was explained through determining the inhibition potential of glucoaurantio-obtusin and aurantio-obtusin towards the glucuronidation metabolism of SN-38 that has been regarded to be the major active product responsible for the toxicity of irinotecan. The results showed that aurantio-obtusin exhibited strong competitive inhibition towards the glucuronidation of SN-38, but negligible inhibition potential of glucoaurantio-obtusin towards SN-38 glucuronidation was observed. These results showed that biotransformation of glucoaurantio-obtusin towards aurantio-obtusin increased the toxicity of irinotecan through increased inhibition of SN-38 glucuronidation.