EMAIL THIS PAGE TO A FRIEND
Antonie van Leeuwenhoek
Nocardioides soli sp. nov., a bacterium isolated from a mountain soil.
PMID 24846051
Abstract
Strain SR-1(T), a Gram-positive, strictly-aerobic, short-rod shaped, non-motile bacterium, was isolated from a mountain soil collected in Seoul Women's University in South Korea. Growth occurred between 15 and 37xa0°C (optimum, 30xa0°C), at pH 6.0-9.0 (optimum, pH 7.0) and with 0-2xa0% NaCl. Phylogenetic analysis based on 16S rRNA gene sequences indicated that strain SR-1(T) belongs to the genus Nocardioides and is closely related to Nocardioides simplex KCTC 9106(T) (96.8xa0%), Nocardioides caeni MN8(T) (96.7xa0%), Nocardioides aromaticivorans H-1(T) (96.6xa0%), and Nocardioides kongijuensis A2-4(T) (96.6xa0%). Chemotaxonomic data revealed that strain SR-1(T) possesses MK-8(H4) as predominant menaquinone, LL-2,6-diaminopimelic acid as the diagnostic diamino acid, phosphatidylglycerol and diphosphatidylglycerol as predominant polar lipids and iso-C16:0, 10-methyl-C18:0, and C18:1 ω9c are major fatty acids. The DNA G+C content of the strain SR-1(T) was 72.4xa0mol%. Based on polyphasic evidence, strain SR-1(T) (=xa0KEMC 9004-134(T)xa0=xa0JCM 19684(T)) should be classified as the type strain of a novel Nocardioides species, for which the name Nocardioides soli sp. nov. is proposed.