PloS one
β1,6 GlcNAc branches-modified PTPRT attenuates its activity and promotes cell migration by STAT3 pathway.
PMID 24846175
Abstract
Receptor-like protein tyrosine phosphatases (RPTPs) are type I transmembrane glycoproteins with N-glycans whose catalytic activities are regulated by dimerization. However, the intrinsic mechanism involved in dimerizing processes remains obscure. In this study, receptor protein tyrosine phosphatase rho (PTPRT) is identified as a novel substrate of N-Acetylglucosaminyltransferase V (GnT-V). We show that addition of β1,6 GlcNAc branches on PTPRT prolongs PTPRT's cell-surface retention time. GnT-V overexpression enhances galectin-3's cell-surface retention and promotes PTPRT's dimerization mediated by galectin-3. Increased dimerization subsequently reduces PTPRT's catalytic activity on the dephosphorylation of signal transducer and activator of transcription 3 (STAT3) at tyrosine residue 705 (pY705 STAT3), then the accumulated pY705 STAT3 translocates into the nucleus. Collectively, these findings provide an insight into the molecular mechanism by which GnT-V promotes cell migration, suggesting that accumulation of β1,6 GlcNAc branched N-glycans promotes PTPRT's dimerization and decreases its catalytic activity, resulting in enhanced cell migratory capacity.