EMAIL THIS PAGE TO A FRIEND
Nature communications
NRBF2 regulates autophagy and prevents liver injury by modulating Atg14L-linked phosphatidylinositol-3 kinase III activity.
PMID 24849286
Abstract
The Beclin 1-Vps34 complex, the core component of the class III phosphatidylinositol-3 kinase (PI3K-III), binds Atg14L or UVRAG to control different steps of autophagy. However, the mechanism underlying the control of PI3K-III activity remains elusive. Here we report the identification of NRBF2 as a component in the specific PI3K-III complex and a modulator of PI3K-III activity. Through its microtubule interaction and trafficking (MIT) domain, NRBF2 binds Atg14L directly and enhances Atg14L-linked Vps34 kinase activity and autophagy induction. NRBF2-deficient cells exhibit enhanced vulnerability to endoplasmic reticulum (ER) stress that is reversed by re-introducing exogenous NRBF2. NRBF2-deficient mice develop focal liver necrosis and ductular reaction, accompanied by impaired Atg14L-linked Vps34 activity and autophagy, although the mice show no increased mortality. Our data reveal a key role for NRBF2 in the assembly of the specific Atg14L-Beclin 1-Vps34-Vps15 complex for autophagy induction. Thus, NRBF2 modulates autophagy via regulation of PI3K-III and prevents ER stress-mediated cytotoxicity and liver injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA001302
Anti-CASP8 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005688
Anti-CASP8 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21154 Anti-Caspase 8 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500404
Anti-Caspase-8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1400524
Anti-NRBF2 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA059477
Anti-NRBF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA021670
Anti-NRBF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501357
Anti-NRBF2 antibody produced in rabbit, affinity isolated antibody
SAB2106597
Anti-NRBF2 antibody produced in rabbit, affinity isolated antibody
K3141
Anti-PIK3C3 (841-853) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406260
Anti-PIK3C3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA040718
Anti-PIK3C3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406748
Anti-SQSTM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB3500430
Anti-SQSTM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104334
Anti-SQSTM1 antibody produced in rabbit, affinity isolated antibody
V9764
Anti-VPS34 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4200399
Anti-WIPI-2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA021488
Anti-WIPI2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA019852
Anti-WIPI2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB3500594
Anti-ZIP3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
EHU123441 MISSION® esiRNA, esiRNA human NRBF2 (esiRNA1)
EHU132031 MISSION® esiRNA, esiRNA human KIAA0831 (esiRNA1)
EMU011921 MISSION® esiRNA, esiRNA targeting mouse D14Ertd436e (esiRNA1)
EMU149141 MISSION® esiRNA, esiRNA targeting mouse Nrbf2
SAB5300353 Monoclonal Anti-CASP8 antibody produced in mouse, clone 1H11, ascites fluid
C4106 Monoclonal Anti-Caspase 8 antibody produced in mouse, ~2 mg/mL, clone CAS8, purified immunoglobulin, buffered aqueous solution
WH0008878M1
Monoclonal Anti-SQSTM1 antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution
SAB1404647
Monoclonal Anti-SQSTM1 antibody produced in mouse, clone 1C9, purified immunoglobulin, buffered aqueous solution