EMAIL THIS PAGE TO A FRIEND
British journal of cancer
High level of HSF1 associates with aggressive endometrial carcinoma and suggests potential for HSP90 inhibitors.
PMID 24853175
Abstract
Recent identification of a specific role of HSF1 in cancer progression has led to new relevance of HSF1 as both a prognostic and a predictive marker. The role of HSF1 in endometrial cancer has so far been unexplored. A total of 823 lesions from endometrial carcinoma precursors, primary tumours and metastases were prospectively collected and explored for HSF1 protein expression in relation to established markers for aggressive disease and survival. Transcriptional alterations related to HSF1 protein level were investigated by microarray analysis for 224 freshly frozen samples in parallel. High expression of HSF1 protein in endometrial carcinoma is significantly associated with aggressive disease and poor survival (all P-values ≤ 0.02), also among ERα-positive patients presumed to have good prognosis. The HSF1-related gene signatures increase during disease progression and were also found to have prognostic value. Gene expression analyses identified HSP90 inhibition as a potential novel therapeutic approach for cases with high protein expression of HSF1. We demonstrate for the first time in endometrial cancer that high expression of HSF1 and measures for transcriptional activation of HSF1 associate with poor outcome and disease progression. The HSP90 inhibitors are suggested as new targeted therapeutics for patients with high HSF1 levels in tumour in particular.