EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
An immortalized human liver endothelial sinusoidal cell line for the study of the pathobiology of the liver endothelium.
PMID 24853805
Abstract
The endothelium lines blood and lymph vessels and protects underlying tissues against external agents such as viruses, bacteria and parasites. Yet, microbes and particularly viruses have developed sophisticated ways to bypass the endothelium in order to gain access to inner organs. De novo infection of the liver parenchyma by many viruses and notably hepatitis viruses, is thought to occur through recruitment of virions on the sinusoidal endothelial surface and subsequent transfer to the epithelium. Furthermore, the liver endothelium undergoes profound changes with age and in inflammation or infection. However, primary human liver sinusoidal endothelial cells (LSECs) are difficult to obtain due to scarcity of liver resections. Relevant derived cell lines are needed in order to analyze in a standardized fashion the transfer of pathogens across the liver endothelium. By lentiviral transduction with hTERT only, we have immortalized human LSECs isolated from a hereditary hemorrhagic telangiectasia (HHT) patient and established the non-transformed cell line TRP3. TRP3 express mesenchymal, endothelial and liver sinusoidal markers. Functional assessment of TRP3 cells demonstrated a high capacity of endocytosis, tube formation and reactivity to immune stimulation. However, TRP3 displayed few fenestrae and expressed C-type lectins intracellularly. All these findings were confirmed in the original primary LSECs from which TRP3 were derived suggesting that these features were already present in the liver donor. We consider TRP3 as a model to investigate the functionality of the liver endothelium in hepatic inflammation in infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1407409
Anti-CD209 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4300650
Anti-CD209 antibody produced in rabbit, affinity isolated antibody
SAB1410626
Anti-CD209 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA007234
Anti-CD81 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500454
Anti-CD81 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS2347
Anti-DC-SIGN (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS2349
Anti-DC-SIGN (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1405802
Anti-FCGR2B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA014730
Anti-FCGR2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410376
Anti-FCGR2B antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2101409
Anti-LYVE1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4300606
Anti-LYVE1 antibody produced in rabbit, affinity isolated antibody
SAB3500048
Anti-SCARB1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700477
Anti-SCARB1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107764
Anti-SCARB1 antibody produced in rabbit, affinity isolated antibody
SAB1300632
Anti-XLKD1 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1307078
ANTI-XLKD1(N-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1403221
Monoclonal Anti-CD209 antibody produced in mouse, clone 3B10, purified immunoglobulin, buffered aqueous solution
SAB4700745 Monoclonal Anti-CD209 antibody produced in mouse, clone UW60.1, purified immunoglobulin, buffered aqueous solution
WH0000975M1
Monoclonal Anti-CD81 antibody produced in mouse, clone 2B7, purified immunoglobulin, buffered aqueous solution
SAB4700232 Monoclonal Anti-CD81 antibody produced in mouse, clone M38, purified immunoglobulin, buffered aqueous solution
D2191
Monoclonal Anti-DC-SIGN1 antibody produced in mouse, clone 120507, purified immunoglobulin, lyophilized powder
WH0002213M1
Monoclonal Anti-FCGR2B antibody produced in mouse, clone 2E10, purified immunoglobulin, buffered aqueous solution
SAB1403809
Monoclonal Anti-FCGR2B antibody produced in mouse, clone 2E10, ascites fluid
SAB1408994 Monoclonal Anti-FCGR2B antibody produced in mouse, clone 1A9, ascites fluid, solution