EMAIL THIS PAGE TO A FRIEND
Cell calcium
Spontaneous calcium transients manifest in the regenerating muscle and are necessary for skeletal muscle replenishment.
PMID 24854233
Abstract
Tissue regeneration entails replenishing of damaged cells, appropriate cell differentiation and inclusion of regenerated cells into functioning tissues. In adult humans, the capacity of the injured spinal cord and muscle to self-repair is limited. In contrast, the amphibian larva can regenerate its tail after amputation with complete recovery of muscle, notochord and spinal cord. The cellular and molecular mechanisms underlying this phenomenon are still unclear. Here we show that upon injury muscle cell precursors exhibit Ca(2+) transients that depend on Ca(2+) release from ryanodine receptor-operated stores. Blockade of these transients impairs muscle regeneration. Furthermore, inhibiting Ca(2+) transients in the regenerating tail prevents the activation and proliferation of muscle satellite cells, which results in deficient muscle replenishment. These findings suggest that Ca(2+)-mediated activity is critical for the early stages of muscle regeneration, which may lead to developing effective therapies for tissue repair.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1106
Ryanodine, ≥98% (HPLC)
C25H35NO9