EMAIL THIS PAGE TO A FRIEND
Biology of reproduction
Galectin-3 contributes to luteolysis by binding to Beta 1 integrin in the bovine corpus luteum.
PMID 24855102
Abstract
Luteolysis is characterized by a reduction in progesterone (P4) production and tissue degeneration in the corpus luteum (CL). One of major events during luteolysis is luteal cell death. Galectin-3, a ubiquitously expressed protein involved in many cellular processes, serves as an antiapoptotic and/or proapoptotic factor in various cell types. Although galectin-3 is detected in the bovine CL, its role remains unclear. The expression of galectin-3 in the bovine CL was higher at the regressed stage than at the other luteal stages. Galectin-3 was localized on luteal steroidogenic cells (LSCs). When cultured LSCs were exposed to prostaglandin F2alpha (PGF) for 48 h, the expression and secretion of galectin-3 increased. When the cultured LSCs were treated with galectin-3 for 24 h, cleaved caspase-3 expression was increased, and the cell viability was decreased, whereas P4 production did not change. Beta 1 integrin, a target protein of galectin-3, was expressed in bovine CL and possessed glycans, which galectin-3 binds. Furthermore, galectin-3 bound to glycans of luteal beta 1 integrin. The decreased cell viability of cultured LSCs by galectin-3 was suppressed by beta 1 integrin antibody. The overall findings suggest that the secreted galectin-3 stimulated by PGF plays a role in structural luteolysis by binding to beta 1 integrin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501430
Anti-Lgals3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406081
Anti-LGALS3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003162
Anti-LGALS3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101338
Anti-LGALS3 antibody produced in rabbit, affinity isolated antibody
SAB1410755
Anti-LGALS3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
D8900 Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
D2906 Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate and phenol red, powder, suitable for cell culture
G5170 Galectin-3 human, recombinant, expressed in E. coli, lyophilized powder
WH0003958M1
Monoclonal Anti-LGALS3 antibody produced in mouse, clone 3C2-2A3, purified immunoglobulin, buffered aqueous solution
R1008 Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
R2655
Monoclonal Anti-Rabbit IgG, Native antibody produced in mouse, ~1 mg/mL, clone RabT-50, purified immunoglobulin, buffered aqueous solution
R3155 Monoclonal Anti-Rabbit IgG, Native−Peroxidase antibody produced in mouse, clone RabT-50, purified from hybridoma cell culture
P3532
Phenol Red, powder, BioReagent, suitable for cell culture
C19H14O5S
114529
Phenol Red, ACS reagent
C19H14O5S
S9133
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Na2O3Se
71950
Sodium selenite, anhydrous, ≥95.0% (RT)
Na2O3Se
214485
Sodium selenite, 99%
Na2O3Se
688770
tert-Butyl acetoacetate, Lonza quality, ≥98.0% (w/w) (GC)
C8H14O3
10911
tert-Butyl acetoacetate, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (GC)
C8H14O3
537594
tert-Butyl acetoacetate, reagent grade, 98%
C8H14O3
B88608
tert-Butyl acetoacetate, reagent grade, 97%
C8H14O3