EMAIL THIS PAGE TO A FRIEND
Oncology reports
HGF and the regulation of tight junctions in human prostate cancer cells.
PMID 24858137
Abstract
Hepatocyte growth factor (HGF) may impact the metastasis of prostate cancer via its action on prostate stem cells or their progeny. Tight junctions (TJs) are crucial to the process of metastasis and have been previously shown to be regulated by HGF. The present study aimed to evaluate the effect of HGF on the function of TJs in human prostate epithelial, prostate stem cell-like and prostate cancer cell lines. Four human prostate cancer cell lines (PC-3, DU-145, PZHPV-7, CaHPV-10), normal adult prostate parental epithelial cells (RWPE-1) and a stem cell-like derivative of RWPE-1 (WPE-STEM) were used to assess HGF-induced changes in TJs. A significant difference was noted in the behaviour between the WPE-STEM, RWPE-1 and the cancer cell lines which was HGF concentration-dependent. However, in the WPE-STEM cells, the effect was biphasic, with the cells seemingly resistant to HGF-modulated TJ disruption. Closer examination revealed that HGF affected the redistribution of ZO-1, ZO-2 and ZO-3 away from the TJs of confluent cells with concurrent loss of claudin-1 and claudin-5, and western blot analysis revealed a loss in TJ protein expression of ZO-1 and ZO-2. We demonstrated for the first time that HGF regulates TJ function in human prostate cells. Moreover, this regulation was dependent on the tumourigenicity of the cells, with the most aggressive cells most susceptible and the stem cell-like cells least susceptible. These data offer an intriguing glimpse of how TJs affect the behaviour of prostate cancer cells and how HGF modulates the expression and function of the molecules maintaining TJ structure and function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA042215
Anti-BRI3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200537
Anti-Claudin-5 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB1406776
Anti-CLDN1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100442
Anti-CLDN5 antibody produced in rabbit, affinity isolated antibody
SAB2100443
Anti-CLDN7 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100444
Anti-CLDN7 (ab2) antibody produced in rabbit, affinity isolated antibody
AV34225
Anti-MLLT4 antibody produced in rabbit, IgG fraction of antiserum
HPA030212
Anti-MLLT4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA030213
Anti-MLLT4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA030214
Anti-MLLT4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
HPA030215
Anti-MLLT4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab4
SAB2102435
Anti-TJP2 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2102436
Anti-TJP2 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1412778
Anti-TJP2 antibody produced in mouse, purified immunoglobulin, 0.5 mg/mL
HPA001813
Anti-TJP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1302474
ANTI-TJP3 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
WH0009076M1
Monoclonal Anti-CLDN1 antibody produced in mouse, clone 1C5-D9, purified immunoglobulin, buffered aqueous solution
SAB1404666
Monoclonal Anti-CLDN1 antibody produced in mouse, clone 2E2-H5, purified immunoglobulin, buffered aqueous solution
WH0007122M1
Monoclonal Anti-CLDN5 antibody produced in mouse, clone 3D8, purified immunoglobulin, buffered aqueous solution
SAB1412779
Monoclonal Anti-TJP2 antibody produced in mouse, clone 4D12, purified immunoglobulin, 0.5 mg/mL